Skip to Content
Merck
All Photos(2)

Key Documents

348414

Sigma-Aldrich

1-Butylimidazole

98%

Synonym(s):

1-Butyl-1H-imidazole, 1-n-Butylimidazole, N-(n-Butyl)imidazole, N-Butyl-1H-imidazole, N-Butylimidazole, N-Butylimidazolium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12N2
CAS Number:
Molecular Weight:
124.18
Beilstein:
109499
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.480 (lit.)

bp

114-116 °C/12 mmHg (lit.)

density

0.945 g/mL at 25 °C (lit.)

SMILES string

CCCCn1ccnc1

InChI

1S/C7H12N2/c1-2-3-5-9-6-4-8-7-9/h4,6-7H,2-3,5H2,1H3

InChI key

MCMFEZDRQOJKMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Butylimidazole acts as an N-coordinated ligand. 1-Butylimidazole is formed as intermediate during the synthesis of a cationic cyclodextrin, used in the chiral separation of amino acids and anionic pharmaceuticals by capillary electrophoresis. Influence of 1-butylimidazole as base on the synthesis of dissymmetric chlorohydrin ester has been reported.

Application

1-Butylimidazole may be used in the synthesis of a heterocyclic mesomeric betaine. It was used in the synthesis of 1-(1-butyl-3-imidazolio)propane-3-sulfonate (BIm3S).

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

293.0 °F

Flash Point(C)

145 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A study of intravascular platelet aggregation by continuous platelet counting.
G M Smith et al.
Thrombosis research, 23(3), 275-288 (1981-08-01)
Viktoriya Polevaya et al.
Materials (Basel, Switzerland), 12(17) (2019-08-31)
The work is devoted to the chemical modification of a polymer that is promising for the creation of gas separation membranes, aimed at increasing the selectivity with respect to CO2. The introduction of ionic liquids into the structure of poly(1-trimethylsilyl-1-propyne)
Nina Gonsior et al.
Beilstein journal of organic chemistry, 8, 390-397 (2012-04-18)
The synthesis of a heterocyclic mesomeric betaine by quaternization reaction of 1-butylimidazole and tetrabromo-1,4-benzoquinone is presented. The structure was verified by means of X-ray single-crystal analysis, NMR and IR spectroscopy. Inclusion complexes of the heterocyclic mesomeric betaine with randomly methylated
A Crockard et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 2(1), 67-72 (1982-01-01)
Autoregulation of cerebral blood flow (CBF) to mean arterial blood pressure (MABP) of 40-50 mm Hg has been demonstrated in the spontaneously breathing gerbil anaesthetised with barbiturate (60 mg/kg). CO2 reactivity has also been assessed at 2.8% change CBF/mm Hg
Carmen Solarte et al.
Molecules (Basel, Switzerland), 16(3), 2065-2074 (2011-03-04)
The anticipated worldwide increase in biodiesel production will result in an accumulation of glycerol for which there are insufficient conventional uses. The surplus of this by-product has increased rapidly during the last decade, prompting a search for new glycerol applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service