Skip to Content
Merck
All Photos(1)

Key Documents

270865

Sigma-Aldrich

Benzene-1,2-dithiol

96%

Synonym(s):

1,2-Dimercaptobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(SH)2
CAS Number:
Molecular Weight:
142.24
Beilstein:
636154
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%
96%

form

liquid

bp

119-120 °C/17 mmHg (lit.)

mp

22-24 °C (lit.)

solubility

water: slightly soluble

density

1.236 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Sc1ccccc1S

InChI

1S/C6H6S2/c7-5-3-1-2-4-6(5)8/h1-4,7-8H

InChI key

JRNVQLOKVMWBFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The transition metal complexes with benzene-1,2-dithiol as ligand were studied by UV-vis, resonance Raman (rR) and infrared (IR) spectroscopies.

Application

Benzene-1,2-dithiol was used in the preparation of a new type of Schiff base that was required in the synthesis of a new series of copper(II) and zinc(II) complexes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lee M, et al.
Royal Society of Chemistry Advances, 5, 86402-86406 (2015)
Taras Petrenko et al.
Journal of the American Chemical Society, 128(13), 4422-4436 (2006-03-30)
Transition metal complexes involving the benzene-1,2-dithiol (L(2-)) and Sellmann's 3,5-di-tert-butylbenzene-1,2-dithiol(L(Bu 2-)) ligands have been studied by UV-vis, infrared (IR), and resonance Raman (rR) spectroscopies. Raman spectra were obtained in resonance with the intervalence charge transfer (IVCT) bands in the near-infrared
N Raman et al.
European journal of medicinal chemistry, 45(11), 5438-5451 (2010-09-25)
A new series of copper(II) and zinc(II) complexes have been designed and synthesized using a new type of Schiff bases derived from the reaction of 3-(3-phenyl-allylidene)-pentane-2,4-dione with para substituted aniline and benzene-1,2-dithiol. Their structures have been established by analytical and
Kallol Ray et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(10), 2783-2797 (2007-02-10)
The electronic structures of [M(L(Bu))(2)](-) (L(Bu)=3,5-di-tert-butyl-1,2-benzenedithiol; M=Ni, Pd, Pt, Cu, Co, Au) complexes and their electrochemically generated oxidized and reduced forms have been investigated by using sulfur K-edge as well as metal K- and L-edge X-ray absorption spectroscopy. The electronic
Masuma Zawari et al.
Antioxidants (Basel, Switzerland), 8(9) (2019-09-05)
Broccoli sprout powder is a rich source of glucosinolates, which are hydrolysed to isothiocyanates in the presence of the enzyme myrosinase. We showed that in vitro incubation of broccoli sprout powder extract with isolated lymphocytes resulted in the upregulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service