Skip to Content
Merck
All Photos(1)

Documents

268992

Sigma-Aldrich

4-Pentyn-2-ol

≥98%

Synonym(s):

(±)-4-Pentyn-2-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CCH2CH(OH)CH3
CAS Number:
Molecular Weight:
84.12
Beilstein:
1734245
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

liquid

refractive index

n20/D 1.438 (lit.)

bp

126-127 °C (lit.)

density

0.892 g/mL at 25 °C (lit.)

SMILES string

CC(O)CC#C

InChI

1S/C5H8O/c1-3-4-5(2)6/h1,5-6H,4H2,2H3

InChI key

JTHLRRZARWSHBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4-Pentyn-2-ol, a secondary homopropargylic alcohol, is toxic with LC50 value of 160.

Application

4-Pentyn-2-ol was used in the synthesis of new Fischer-type rhenium (III) benzoyldiazenido-2-oxacyclocarbenes. It was also used in the preparation of c-aryl glycosides by undergoing cycloaddition reaction with an aryl nitrile oxide.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorenza Marvelli et al.
Dalton transactions (Cambridge, England : 2003), (5)(5), 713-722 (2004-07-15)
A family of new Fischer-type rhenium(III) benzoyldiazenido-2-oxacyclocarbenes of formula [(ReCl2[eta1-N2C(O)Ph][=C(CH2)nCH(R)O](PPh3)2][n = 2, R = H (2), R = Me (3); n = 3, R = H (4), R = Me (5)] have been prepared by reaction of [ReCl2[eta2-N2C(Ph)O](PPh3)2] (1) with
G D Veith et al.
Xenobiotica; the fate of foreign compounds in biological systems, 19(5), 555-565 (1989-05-01)
1. The 96-h LC50 values for 16 acetylenic alcohols in the fathead minnow (Pimephales promelas) were determined using continuous-flow diluters. The measured LC50 values for seven tertiary propargylic alcohols agreed closely with the QSAR predictions based upon data for other
Frank M Hauser et al.
Organic letters, 4(6), 977-978 (2002-03-15)
[reaction: see text] A six-step route for de novo synthesis of C-aryl glycosides based on cycloaddition of an aryl nitrile oxide with 4-pentyn-2-ol has been developed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service