Skip to Content
Merck
All Photos(2)

Documents

130850

Sigma-Aldrich

3-Butyn-1-ol

97%

Synonym(s):

3-Butynol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CCH2CH2OH
CAS Number:
Molecular Weight:
70.09
Beilstein:
773710
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39020335
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.441 (lit.)

bp

128.9 °C (lit.)

mp

−63.6 °C (lit.)

density

0.926 g/mL at 25 °C (lit.)

SMILES string

OCCC#C

InChI

1S/C4H6O/c1-2-3-4-5/h1,5H,3-4H2

InChI key

OTJZCIYGRUNXTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The enthalpies of combustion and vaporization of 3-butyn-1-ol is measured by static bomb combustion calorimetry and correlation gas chromatography techniques.

Application

3-Butyn-1-ol was used to prepare Markovnikov addition product.
Alkynyl substrate used in a study of a palladium-catalyzed coupling with β-tetrionic acid bromide leading to alkynyl substituted furanones in good yield.
Synthon for preparation of oxygen-containing heterocycles and protected esters of s-Hydroxy-L-isoleucine

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kopf, Henning; Pietraszuk. et al.
Organometallics, 25(10), 2533-2546 (2006)
Deniz Dogruer et al.
Bioorganic & medicinal chemistry letters, 14(2), 523-526 (2003-12-31)
The binding of a series pyridylbutynylamines 6 was examined at alpha4beta2 nACh receptors. Structural modifications, comparing 6 with pyridyl ethers 2, did not consistently result in parallel effects on receptor affinity, suggesting possible differences in their modes of binding. Furthermore
Sumod A Pullarkat et al.
Inorganic chemistry, 45(18), 7455-7463 (2006-08-29)
Alcohol functionalized diphosphine ligands with chirality residing on the carbon backbone were prepared using a novel two-stage asymmetric synthetic methodology from the corresponding terminal alkynols. Under mild conditions, the alkynols, 3-butyn-1-ol and 2-propyn-1-ol, were subjected to direct hydrophosphination to give
Kristen Bennett et al.
Applied and environmental microbiology, 82(8), 2270-2279 (2016-01-31)
Nitrosomonas europaea is an aerobic nitrifying bacterium that oxidizes ammonia (NH3) to nitrite (NO2 (-)) through the sequential activities of ammonia monooxygenase (AMO) and hydroxylamine dehydrogenase (HAO). Many alkynes are mechanism-based inactivators of AMO, and here we describe an activity-based
Ederley Vélez et al.
The journal of physical chemistry. A, 109(34), 7832-7838 (2006-07-13)
The enthalpies of combustion and vaporization of 3-buten-1-ol and 3-butyn-1-ol have been measured by static bomb combustion calorimetry and correlation gas chromatography techniques, respectively, and the gas-phase enthalpies of formation, Delta(f)H degrees (m)(g), have been determined, the values being -147.3

Articles

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service