Skip to Content
Merck
All Photos(2)

Key Documents

265772

Sigma-Aldrich

Gold

powder, <45 μm, 99.99% trace metals basis

Synonym(s):

Gold Powder, Gold element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Au
CAS Number:
Molecular Weight:
196.97
EC Number:
MDL number:
UNSPSC Code:
12141717
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99.99% trace metals basis

form

powder

resistivity

2.05 μΩ-cm, 0°C

particle size

<45 μm

bp

2808 °C (lit.)

mp

1063 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

General description

Gold (Au) is a yellow noble metal known for its lustrous appearance and high resistance to corrosion. With a melting point of 1,063 °C, gold is one of the most malleable and ductile metals, making it ideal for various applications. Our high-purity gold powder is characterized by its purity of 99.995% on a trace metals basis and is available in a fine powder form with particle sizes less than 45 microns. Gold is widely used in electronics, dental materials, and as a catalyst in various chemical reactions. Its exceptional conductivity and biocompatibility make it particularly valuable in electronics, catalysis, and synthesis.[1][2][3]

Application

Applied in the synthesis of new low-dimensional, quaternary gold selenophosphate compounds, A3AuP2Se8 (a=Rb, K, Cs) and A2Au2P2Se6 (a=Rb, K).[4]

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chondroudis, K. et al.
Inorganic Chemistry, 36, 2623-2623 (1997)
Zhengxia Liu et al.
Journal of biomedical nanotechnology, 9(6), 1017-1028 (2013-07-19)
Ischemic stroke is a leading cause of death and disability, and the treatment options are limited. Recent studies demonstrated that the promising applications of gold nanoparticles (Au-NPs) as intrinsic therapeutics; however, little is known about the effect of Au-NPs on
Young Joo Choi et al.
Journal of nanoscience and nanotechnology, 13(6), 4437-4445 (2013-07-19)
Gold nanorods (Au NRs) that absorb near-infrared (NIR) light have great potential in the field of nanomedicine. Photothermal therapy (PTT), a very attractive cancer therapy in nanomedicine, combines nanomaterials and light. The aim of this study was to elucidate the
Matthieu B Miller et al.
Journal of the Air & Waste Management Association (1995), 63(6), 681-693 (2013-07-19)
Industrial gold mining is a significant source of mercury (Hg) emission to the atmosphere. To investigate ways to reduce these emissions, reclamation and dust and mercury control methods used at open pit gold mining operations in Nevada were studied in
Nidhi Gupta et al.
Journal of nanoscience and nanotechnology, 13(7), 4917-4924 (2013-08-02)
The preparation and photocatalytic reductive ability of a new spongy flower or cotton bud like amorphous CdS microstructures (size = 200-300 nm) having intermediate crystal phase between cubic and hexagonal structure and possesses much larger surface area ca. 119 m2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service