Skip to Content
Merck
All Photos(1)

Key Documents

268461

Sigma-Aldrich

Gold

foil, thickness 0.025 mm, 99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Au
CAS Number:
Molecular Weight:
196.97
EC Number:
MDL number:
UNSPSC Code:
12141717
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.99% trace metals basis

form

foil

resistivity

2.05 μΩ-cm, 0°C

thickness

0.025 mm

bp

2808 °C (lit.)

mp

1063 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Related Categories

General description

Gold is one of the most popular materials to be used for neutron flux monitoring mainly because it possesses a large thermal cross section for neutron capture (197Au(η, η) 198Au. Gold has the half-life of 2.7 days. Reports show that the rate of dissolution of Au is very fast in SnPb solder.

Application

Gold based neutron flux monitors may use gold foils. Au foils may be used to form a AuSn/Au joint system for opto-electronic chips. Modified gold foil electrode may be used to study heterogeneous electron transfer properties of biological electron transfer proteins. 3 Electrodeposited polycrystalline palladium-nickel alloy on gold foils may be investigated for the enhanced catalytic behavior of the alloy.

Quantity

300 mg = 25 × 25 mm; 1.2 g = 50 × 50 mm

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Study of wetting reaction between eutectic AuSn and Au foil.
Lai YT and Liu CY
Journal of Electronic Materials, 35, 28-34 (2006)
Performance and comparison of gold-based neutron flux monitors.
Steinhauser G, et al.
Gold Bulletin, 45, 17-22 (2012)
A kinetic study of oxygen reduction reaction on palladium-nickel alloy surfaces.
Li B, et al.
Electrochemical Society Transactions, 6(25), 139-144 (2008)
Zhengxia Liu et al.
Journal of biomedical nanotechnology, 9(6), 1017-1028 (2013-07-19)
Ischemic stroke is a leading cause of death and disability, and the treatment options are limited. Recent studies demonstrated that the promising applications of gold nanoparticles (Au-NPs) as intrinsic therapeutics; however, little is known about the effect of Au-NPs on
Young Joo Choi et al.
Journal of nanoscience and nanotechnology, 13(6), 4437-4445 (2013-07-19)
Gold nanorods (Au NRs) that absorb near-infrared (NIR) light have great potential in the field of nanomedicine. Photothermal therapy (PTT), a very attractive cancer therapy in nanomedicine, combines nanomaterials and light. The aim of this study was to elucidate the

Articles

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service