Skip to Content
Merck
All Photos(1)

Key Documents

260908

Sigma-Aldrich

N-Bromophthalimide

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4BrNO2
CAS Number:
Molecular Weight:
226.03
Beilstein:
131211
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

mp

194-198 °C (lit.)

functional group

imide

SMILES string

BrN1C(=O)c2ccccc2C1=O

InChI

1S/C8H4BrNO2/c9-10-7(11)5-3-1-2-4-6(5)8(10)12/h1-4H

InChI key

MARXMDRWROUXMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Bromophthalimide has been used:
  • as reagent in allylic amination reactions of alkenes
  • brominating reagent in enantioselective synthesis of multisubstituted biaryl derivatives by chiral phosphoric acid catalyzed asymmetric bromination
  • as a titrant in titrimetric determination of isoniazid in pure form or in tablets

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Krishnakumar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 62(4-5), 918-925 (2005-06-14)
Fourier transform infrared (FT-IR) spectra of phthalimide and N-bromophthalimide have been recorded in the range of 4000-400 cm-1. With the hope of providing more and effective information on the fundamental vibrations, a normal coordinate analysis has been performed on phthalimide
Feng Chen et al.
Journal of the American Chemical Society, 135(4), 1232-1235 (2013-01-15)
A catalytic asymmetric bromocyclization of trisubstituted olefinic amides that uses a C(2)-symmetric mannitol-derived cyclic selenium catalyst and a stoichiometric amount of N-bromophthalimide is reported. The resulting enantioenriched pyrrolidine products, which contain two stereogenic centers, can undergo rearrangement to yield 2,3-disubstituted
Titrimetric determination of para-aminobenzoic acid using N-bromophthalimide and N-bromosaccharin.
K G Kumar et al.
Journal of pharmaceutical and biomedical analysis, 7(5), 627-631 (1989-01-01)
Tejas P Pathak et al.
Journal of the American Chemical Society, 134(14), 6120-6123 (2012-04-03)
We report the site-selective bromination of vancomycin to produce, with substantial efficiency, previously unknown monobromovancomycins, a dibromovancomycin, and a tribromovancomycin. We document the inherent reactivity of native vancomycin toward N-bromophthalimide. We then demonstrate significant rate acceleration and perturbation of the
A M el-Brashy et al.
Journal of pharmaceutical and biomedical analysis, 10(6), 421-426 (1992-06-01)
Two methods are proposed for the determination of isoniazid in pure form or in tablets. In the first method chlorpromazine hydrochloride, when treated with 2-iodoxybenzoic acid as an oxidant in 50% w/v o-phosphoric acid solution, is oxidized to chlorpromazine free

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service