Skip to Content
Merck
All Photos(1)

Key Documents

259020

Sigma-Aldrich

Dibromomethane-d2

≥99 atom % D, ≥99% (CP), contains copper as stabilizer

Synonym(s):

Methylene-d2 bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD2Br2
CAS Number:
Molecular Weight:
175.85
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

vapor density

5.9 (vs air)

vapor pressure

34.9 mmHg ( 20 °C)

isotopic purity

≥99 atom % D

Assay

≥99% (CP)

form

liquid

contains

copper as stabilizer

technique(s)

NMR: suitable

refractive index

n20/D 1.5381 (lit.)

bp

99 °C (lit.)

density

2.505 g/mL at 25 °C

mass shift

M+2

SMILES string

[2H]C([2H])(Br)Br

InChI

1S/CH2Br2/c2-1-3/h1H2/i1D2

InChI key

FJBFPHVGVWTDIP-DICFDUPASA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

[18 F] FMeNER-D2: Reliable fully-automated synthesis for visualization of the norepinephrine transporter.
Rami-Mark C, et al.
Nuclear Medicine and Biology, 40(8), 1049-1054 (2013)
PET evaluation of novel radiofluorinated reboxetine analogs as norepinephrine transporter probes in the monkey brain.
Schou M, et al.
Synapse, 53(2), 57-67 (2004)
Salla Lahdenpohja et al.
Journal of labelled compounds & radiopharmaceuticals, 63(9), 408-418 (2020-05-07)
Here, we describe the development of an in-house-built device for the fully automated multistep synthesis of the cannabinoid CB1 receptor imaging tracer (3R,5R)-5-(3-([18 F]fluoromethoxy-d2 )phenyl)-3-(((R)-1-phenylethyl)amino)-1-(4-(trifluoromethyl)phenyl)pyrrolidin-2-one ([18 F]FMPEP-d2 ), following good manufacturing practices. The device is interfaced to a HPLC and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service