Skip to Content
Merck
All Photos(2)

Key Documents

256064

Sigma-Aldrich

Ammonium nitrate

99.999% trace metals basis

Synonym(s):

Nitric acid ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4NO3
CAS Number:
Molecular Weight:
80.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

crystals and lumps

impurities

≤20.0 ppm Trace Metal Analysis

bp

210 °C (lit.)

mp

169 °C (lit.)

SMILES string

N.O[N+]([O-])=O

InChI

1S/HNO3.H3N/c2-1(3)4;/h(H,2,3,4);1H3

InChI key

PRORZGWHZXZQMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Synthesis of 3-Amino-4-substituted Monocyclic ß-Lactams-Important Structural Motifs in Medicinal Chemistry.: This study presents a novel synthesis route for 3-amino-4-substituted monocyclic ß-lactams, which are significant in medicinal chemistry due to their biological activities. The process utilizes ammonium nitrate as a reagent for the synthesis, highlighting its role in advanced organic chemistry applications (Grabrijan et al., 2021).

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1C - Ammonium nitrate and ammonium nitrate containing preparations

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sari Palmroth et al.
Annals of botany, 111(3), 467-477 (2013-01-10)
Water and nitrogen (N) are two limiting resources for biomass production of terrestrial vegetation. Water losses in transpiration (E) can be decreased by reducing leaf stomatal conductance (g(s)) at the expense of lowering CO(2) uptake (A), resulting in increased water-use
Deborah Wakeham et al.
Physical chemistry chemical physics : PCCP, 14(15), 5106-5114 (2012-01-31)
The nanostructure of the ethanolammonium nitrate (EtAN)-air surface has been investigated using X-ray reflectometry (XRR), vibrational sum frequency spectroscopy (VSFS) and neutral impact collision ion scattering spectroscopy (NICISS). The XRR data decays more rapidly than expected for a perfectly sharp
Mohammad Mahdi Najafpour et al.
Dalton transactions (Cambridge, England : 2003), 41(34), 10156-10160 (2012-07-27)
Nanoscale manganese oxides within Faujasite zeolite have been synthesized with a simple method and characterized by scanning electron microscopy, X-ray diffraction spectrometry, N(2) adsorption-desorption isotherms, transmission electron microscopy, and atomic absorption spectroscopy. These oxides showed efficient water oxidizing activity in
Elaine Ward et al.
PloS one, 7(4), e35657-e35657 (2012-05-05)
The alkaline serine protease VCP1 of the fungus Pochonia chlamydosporia belongs to a family of subtilisin-like enzymes that are involved in infection of nematode and insect hosts. It is involved early in the infection process, removing the outer proteinaceous vitelline
Padmakar A Suryavanshi et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(39), 13207-13215 (2013-08-16)
The cerium(IV) ammonium nitrate (CAN)-catalyzed sequential multicomponent reaction between tryptamine, α,β-unsaturated aldehydes, and β-dicarbonyl compounds affords highly substituted indolo[2,3-a]quinolizines in a single synthetic operation. Two rings are generated through the creation of two C-C and two C-N bonds by a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service