Skip to Content
Merck

07592

trans-2-Nonenal

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)5CH=CHCHO
CAS Number:
Molecular Weight:
140.22
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
85151701
EC Number:
242-609-6
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

vapor density

>1 (vs air)

assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

refractive index

n20/D 1.453 (lit.), n20/D 1.454-1.458

bp

88-90 °C/12 mmHg (lit.)

density

0.846 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[H]C(=O)C(\[H])=C(/[H])CCCCCC

InChI

1S/C9H16O/c1-2-3-4-5-6-7-8-9-10/h7-9H,2-6H2,1H3/b8-7+

InChI key

BSAIUMLZVGUGKX-BQYQJAHWSA-N

General description

trans-2-Nonenal (T2N) is a volatile unsaturated aldehyde with an unpleasant greasy odor, present in low concentrations in barley, malt, and certain vegetables. It is utilized as an insect repellent, insecticide, and flavor compound. T2N is one of the basic components contributing to the off-flavor and odor in stored beer.

Application

trans-2-Nonenal (T2N) may be used as a reference standard for the determination of the analyte in barley and malt samples by analytical methods.


Still not finding the right product?

Explore all of our products under trans-2-Nonenal


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

10 - Combustible liquids

flash_point_f

183.2 °F - closed cup

flash_point_c

84 °C - closed cup

target_organs

Respiratory system



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library