254282
Lithium metaborate
99.995% trace metals basis
Synonym(s):
Boric acid lithium salt
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
LiBO2
CAS Number:
Molecular Weight:
49.75
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder
Recommended Products
grade
for analytical purposes
Assay
99.995% trace metals basis
form
powder
impurities
≤55.0 ppm Trace Metal Analysis
mp
845 °C (lit.)
SMILES string
[Li+].[O-]B=O
InChI
1S/BO2.Li/c2-1-3;/q-1;+1
InChI key
HZRMTWQRDMYLNW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
T Vilaithong et al.
Radiation protection dosimetry, 100(1-4), 211-216 (2002-10-18)
High resolution emission spectra of several TL materials, that are commonly used in dosimetry, were measured using a low cost fibre optic spectrometer containing a 2048 pixel CCD array. The spectra were taken from 177 to 890 nm with a
Josselin Morand et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 67(7-8 Suppl), S149-S152 (2009-04-21)
This work provides the basis of a methodology to build a deterministic model for the spatial distribution of the (10)B(n,alpha)(7)Li reaction rate in boron neutron capture therapy (BNCT), as a function of space variables, boron concentration and beam incidence angle
Zheng Quan et al.
Applied optics, 48(16), 2979-2982 (2009-06-03)
Efficient cw intracavity frequency doubling of a diode end-pumped Nd:GdVO4 laser that operates in the 4F(3/2) --> 4I(9/2) transition at 912 nm is demonstrated. A 15 mm long lithium borate crystal, cut for critical type I phase matching at room
P E Warwick et al.
Analytical chemistry, 73(14), 3410-3416 (2001-07-31)
Americium-241 and plutonium determinations will become of greater importance over the coming decades as 137Cs and 241Pu decay. The impact of 137Cs on environmental chronology has been great, but its potency is waning as it decays and diffuses. Having 241Am
Woo Jin Ko et al.
Journal of endourology, 26(4), 313-317 (2012-03-24)
The goal of this study is to identify the most efficient sweeping angle (SA) during photoselective vaporization of the prostate (PVP). Experiments were conducted with GreenLight XPS™ laser at 120 and 180 W. Ten blocks of porcine kidney were used for
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service