253340
3-Quinuclidinol
99%
Synonym(s):
1-Azabicyclo[2.2.2]octan-3-ol, 3-Hydroxyquinuclidine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C7H13NO
CAS Number:
Molecular Weight:
127.18
Beilstein:
80122
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
mp
220-223 °C (lit.)
functional group
hydroxyl
SMILES string
OC1CN2CC[C@H]1CC2
InChI
1S/C7H13NO/c9-7-5-8-3-1-6(7)2-4-8/h6-7,9H,1-5H2
InChI key
IVLICPVPXWEGCA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
(R)-3-Quinuclidinol is a valuable compound for the production of various pharmaceuticals.
Application
3-Quinuclidinol has been used:
- as chiral building block for many antimuscarinic agents
- in chemoselective α−iodination of various simple and multi-functionalised acrylic esters via Morita-Baylis-Hillman protocol
- as reagent for cleavage of β-keto and vinylogous β-keto esters
- as synthon for the preparation of cholinergic receptor ligands and anesthetics
- as catalyst for condensation of methyl vinyl ketone with aldehydes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Selective Cleavage of ?-Keto and Vinylogous ?-Keto Esters by 3-Quinuclidinol.
Parish EJ, et al.
Synthetic Communications, 5(5), 341-345 (1975)
Wen-Xia Zhang et al.
Organic letters, 15(19), 4917-4919 (2013-09-21)
A new keto reductase (ArQR), identified from Agrobacterium radiobacter ECU2556, can efficiently reduce 3-quinuclidinone in excellent enantioselectivity and high space-time yield for the synthesis of (R)-3-quinuclidinol, a chiral building block of many antimuscarinic agents. This is the first time that
Tetrahedron Letters, 31, 4509-4509 (1990)
Khim.-Farm. Zh., 26, 12-12 (1992)
Synthesis and local anaesthetic activities of 3-(2-alkoxyphenylcarbamoyloxy)chinuclidinium chlorides.
F Gregán et al.
Die Pharmazie, 48(6), 465-466 (1993-06-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service