Skip to Content
Merck
All Photos(2)

Documents

244627

Sigma-Aldrich

Zinc sulfide

powder, 10 μm, 99.99% trace metals basis

Synonym(s):

Zinc sulphide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnS
CAS Number:
Molecular Weight:
97.46
EC Number:
MDL number:
UNSPSC Code:
12161600
eCl@ss:
38150412
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99.99% trace metals basis

form

powder

reaction suitability

core: zinc
reagent type: catalyst

particle size

10 μm

density

4.1 g/mL at 25 °C (lit.)

SMILES string

S=[Zn]

InChI

1S/S.Zn

InChI key

WGPCGCOKHWGKJJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Uses:
  • Preparation of flexible transparent conductive coatings essential for fabrication of a variety of printed electronic devices such as flexible displays and solar cells
  • Prepare a composite CdS-ZnS/Zirconium-titanium phosphate (ZTP) photocatalyst for hydrogen production under visible light
  • Prepare light-controlled bioelectrochemical sensors based on CdSe/ZnS quantum dots
  • Catalyst for photocatalytic degradation of organic pollutants
  • Preparation of color tunable light-emitting diodes (LEDs)
  • Prepare (CdS-ZnS)-TiO2 combined photocatalysts for electricity production via photoelectrocatalysis
  • Catalyst for synthesis of spirooxindole derivatives in aqueous medium via Knoevenagel condensation followed by Michael addition
  • Prepare CdSe/ZnS q uantum dots for chemiluminescent and chemiluminescence resonance energy transfer (CRET) detection of DNA, metal ions, and aptamer-substrate complexes
  • Preparation of ZnS nanocrystals for ultrasensitive protein detection in terms of multiphonon resonance Raman scattering
Zinc sulfide can be used:
  • To prepare flexible transparent conductive coatings essential for fabrication of a variety of printed electronic devices such as flexible displays and solar cells.
  • To prepare a composite CdS-ZnS/zirconium-titanium phosphate (ZTP) photocatalyst for hydrogen production under visible light.
  • To prepare light-controlled bioelectrochemical sensors based on CdSe/ZnS quantum dots.
  • As a catalyst for the photocatalytic degradation of organic pollutants.
  • In the preparation of color tunable light-emitting diodes (LEDs).
  • To prepare (CdS-ZnS)-TiO2 combined photocatalysts for electricity production via photoelectrocatalysis.
  • As a catalyst for the synthesis of spirooxindole derivatives in aqueous medium via Knoevenagel condensation followed by Michael addition.
  • To prepare CdSe/ZnS quantum dots for chemiluminescent and chemiluminescence resonance energy transfer (CRET) detection of DNA, metal ions, and aptamer-substrate complexes.
  • To prepare ZnS nanocrystals for ultrasensitive protein detection in terms of multiphonon resonance Raman scattering.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Ultrasensitive protein detection in terms of multiphonon resonance Raman scattering in ZnS nanocrystals.
Chu X, et al.
Applied Physics Letters, 98(25), 253703-253703 (2011)
Flexible transparent conductive coatings by combining self-assembly with sintering of silver nanoparticles performed at room temperature.
Layani M and Magdassi S
Journal of Materials Chemistry, 21(39), 15378-15382 (2011)
Photocatalysis and photoelectrocatalysis using (CdS-ZnS)/TiO2 combined photocatalysts.
Antoniadou M, et al.
Applied Catalysis. B, Environmental, 107(1-2), 188-196 (2011)
Efficient hydrogen production by composite photocatalyst CdS?ZnS/Zirconium?titanium phosphate (ZTP) under visible light illumination.
Biswal N, et al.
International Journal of Hydrogen Energy, 36(21), 13452-13460 (2011)
Visible-light absorptivity of a zincoxysulfide (ZnOxS1? x) composite semiconductor and its photocatalytic activities for degradation of organic pollutants under visible-light irradiation.
Kim C, et al.
Applied Catalysis A: General, 330, 127-133 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service