Skip to Content
Merck
All Photos(1)

Key Documents

237612

Sigma-Aldrich

(R)-(−)-1,3-Butanediol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2CH2OH
CAS Number:
Molecular Weight:
90.12
Beilstein:
1718944
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.1 (vs air)

vapor pressure

0.06 mmHg ( 20 °C)

Assay

95%

form

liquid

optical activity

[α]21/D −31.0°, c = 1 in ethanol

optical purity

ee: 95% (GLC)

autoignition temp.

741 °F

bp

107-110 °C/23 mmHg (lit.)

density

1.005 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

C[C@@H](O)CCO

InChI

1S/C4H10O2/c1-4(6)2-3-5/h4-6H,2-3H2,1H3/t4-/m1/s1

InChI key

PUPZLCDOIYMWBV-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Q Wu et al.
Guang pu xue yu guang pu fen xi = Guang pu, 20(5), 612-613 (2003-08-30)
In the present paper, we studied the surface structure effect of Pt electrode modified with antimony and sulfur adatoms towards 1,3-Butanediol oxidation. The results demonstrated that the modification of Sb and S both inhibited the dissociative adsorption of 1,3-Butanediol into
Nobuya Itoh et al.
Applied microbiology and biotechnology, 75(6), 1249-1256 (2007-04-20)
An asymmetric hydrogen-transfer biocatalyst consisting of mutated Rhodococcus phenylacetaldehyde reductase (PAR) or Leifsonia alcohol dehydrogenase (LSADH) was applied for some water-soluble ketone substrates. Among them, 4-hydroxy-2-butanone was reduced to (S)/(R)-1,3-butanediol, a useful intermediate for pharmaceuticals, with a high yield and
Biagio Velino et al.
The journal of physical chemistry. A, 115(34), 9585-9589 (2011-03-23)
The rotational spectra of five conformers of 1,3-butandiol have been measured by pulsed jet Fourier transform microwave spectroscopy. All of them are stabilized by an internal hydrogen bond and all of them have a GG' or a G'G arrangement of
Yehong Wang et al.
Journal of the American Chemical Society, 135(4), 1506-1515 (2012-12-12)
The use of a heterogeneous Lewis acid catalyst, which is insoluble and easily separable during the reaction, is a promising option for hydrolysis reactions from both environmental and practical viewpoints. In this study, ceria showed excellent catalytic activity in the
A Baudot et al.
Cryobiology, 40(2), 151-158 (2000-05-02)
The glass-forming tendency on cooling and the stability of the wholly amorphous state on warming have been previously reported for many cryoprotective solutions. However, unlike the other solutions, those of dimethyl sulfoxide (Me(2)SO) have not been studied on cooling. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service