Skip to Content
Merck
All Photos(4)

Key Documents

224758

Sigma-Aldrich

Tris(2,2′-bipyridyl)dichlororuthenium(II) hexahydrate

powder

Synonym(s):

Ru(BPY)3, Ruthenium-tris(2,2′-bipyridyl) dichloride, Tris(2,2′-bipyridyl)ruthenium(II) chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H24Cl2N6Ru · 6H2O
CAS Number:
Molecular Weight:
748.62
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

reaction suitability

core: ruthenium
reaction type: Photocatalysis
reagent type: catalyst

mp

>300 °C (lit.)

photocatalyst activation

450 nm

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Ru]Cl.c1ccc(nc1)-c2ccccn2.c3ccc(nc3)-c4ccccn4.c5ccc(nc5)-c6ccccn6

InChI

1S/3C10H8N2.2ClH.6H2O.Ru/c3*1-3-7-11-9(5-1)10-6-2-4-8-12-10;;;;;;;;;/h3*1-8H;2*1H;6*1H2;/q;;;;;;;;;;;+2/p-2

InChI key

WHELTKFSBJNBMQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Tris(2,2′-bipyridyl)dichlororuthenium(II) hexahydrate is a metal complex and is used as an luminescent dye. The metal complex in its ground state can be excited by visible light which leads to the formation of a spin-allowed excited state. This excited state undergoes radiationless deactivation very quickly to form spin-forbidden, long-lived luminescent excited state. It is used widely in electroluminescent devices.

Application

  • Used in the synthesis of hybrid catalysts which are used in oxidase based biosensors.
  • Used as a luminophore for multiplexed signalling in bio analysis.
  • Used in oxygen assesment in living cells.
Learn More at the Professor and Product Portal of Professor Teshik Yoon and Professor Corey Stephenson.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stéphanie Heux et al.
Applied and environmental microbiology, 77(19), 7040-7049 (2011-08-16)
Miniaturization and high-throughput screening are currently the focus of emerging research areas such as systems biology and systems biotechnology. A fluorescence-based screening assay for the online monitoring of oxygen and pH and a numerical method to mine the resulting online
Old Molecules, New Concepts:[Ru (bpy) 3] 2+ as a Molecular Encoder-Decoder
Ceroni, P., Bergamini, G., & Balzani, V.
Angewandte Chemie (International Edition in English), 121(45), 8668-8670 (2009)
A Theoretical and Experimental Framework for Understanding Electrogenerated Chemiluminescence (ECL) Emission at Bipolar Electrodes
Mavre F, et al
Analytical Chemistry, 81, 6218-6225 (2009)
Attapulgite Clay Combined with Tris(2,2'bipyridyl)ruthenium(II) for the Enhancement of the
Electrogenerated Chemiluminescence Sensing
Gu B. et al
International Journal of Electrochemical Science, 7, 6202-6213 (2012)
Novel organic-inorganic hybrid material based on tris (2, 2'-bipyridyl) dichlororuthenium (II) hexahydrate and Dawson-type tungstophosphate K 7[H4PW18O62]? 18H2O as a bifuctional hydrogen peroxide electrocatalyst for biosensors
Ammam, M., & Easton, E. B.
Sensors and Actuators B, Chemical, 161(1), 520-527 (2012)

Related Content

Research in the Stephenson lab focuses upon the development of new chemical methods which enable the activation of chemical bonds under mild reaction conditions.

One major theme of research in the Yoon lab is the development of unique chemical transformations enabled by visible light photochemistry. A variety of transition metal photocatalysts responsive to visible light are known, and the Yoon lab has shown that their well understood photochemistry can be coupled to cycloadditions, arene oxidations, amine functionalizations, and thiol-ene reactions, among others.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service