Skip to Content
Merck
All Photos(2)

Key Documents

223662

Sigma-Aldrich

Tris(triphenylphosphine)ruthenium(II) dichloride

97%

Synonym(s):

Tris(triphenylphosphine)dichlororuthenium, Dichlorotris(triphenylphosphine)ruthenium(II), Ruthenium(II)-tris(triphenylphosphine) dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)3P]3RuCl2
CAS Number:
Molecular Weight:
958.83
Beilstein:
4935939
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

core: ruthenium
reagent type: catalyst
reaction type: C-H Activation

SMILES string

Cl[Ru]Cl.c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9

InChI

1S/3C18H15P.2ClH.Ru/c3*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h3*1-15H;2*1H;/q;;;;;+2/p-2

InChI key

WIWBLJMBLGWSIN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Tris(triphenylphosphine)ruthenium(II) dichloride or [RuCl2(PPh3)3] can be used as a catalyst to synthesize:
  • Functionalized alcohols by C-C cross-coupling reaction between different alcohols via sp3 C-H bond activation of primary alcohols in the presence of Lewis acid.
  • Furan derivatives from allenyl sulfides via 1,4 migration of the sulfanyl group.
  • 1,3-diphenylpropan-1-one by alkylation of acetophenone with benzyl alcohol via C-C bond formation.
  • Vinyl chloride monomer by hydrochlorination reaction of acetylene.

RuCl2(PPh3)3 can also be used as a catalyst in the cyclization, isomerization, reduction, oxidation, and cross-coupling reactions of a variety of organic products. Hydrogenation of nitro groups, imines, and ketones, as well as selective oxidation of alcohols are also possible in the presence of this catalyst.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dichlorotris (triphenylphosphine) ruthenium (II) a
Plummer JS, et al.
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Cross-coupling reaction between alcohols through sp3 C-H activation catalyzed by a ruthenium/Lewis acid system.
Shu-Yu Zhang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(33), 10201-10205 (2008-10-11)
Transition-Metal-Catalyzed Rearrangement of Allenyl Sulfides: A Route to Furan Derivatives
Peng L, et al.
Angewandte Chemie (International Edition in English), 46(11), 1905-1908 (2007)
Synthesis of vinyl chloride monomer over carbon-Supported tris-(triphenylphosphine) ruthenium dichloride catalysts
Li Xing, et al.
Catalysts, 8(7), 276-276 (2018)
Cross-Coupling Reaction between Alcohols through sp3 C-H Activation Catalyzed by a Ruthenium/Lewis Acid System
Zhang S-Y, et al.
Chemistry?A European Journal , 14(33), 10201-10205 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service