21612
L-(−)-α-Amino-ε-caprolactam hydrochloride
≥97.0% (AT)
Synonym(s):
(S)-3-Amino-hexahydro-2-azepinone hydrochloride, L-Lysine lactam hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H12N2O · HCl
CAS Number:
Molecular Weight:
164.63
Beilstein:
4820243
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥97.0% (AT)
form
solid
optical activity
[α]20/D −27±2°, c = 1% in H2O
optical purity
enantiomeric ratio: ≥99:1 (HPLC)
storage temp.
2-8°C
SMILES string
Cl[H].N[C@H]1CCCCNC1=O
InChI
1S/C6H12N2O.ClH/c7-5-3-1-2-4-8-6(5)9;/h5H,1-4,7H2,(H,8,9);1H/t5-;/m0./s1
InChI key
LWXJCGXAYXXXRU-JEDNCBNOSA-N
Application
L-(−)-α-Amino-ε-caprolactam hydrochloride can be used to introduce caprolactam moiety during the total synthesis of bengamides, which are bioactive compounds naturally found in marine sponges.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Total synthesis of bengamide E and analogues by modification at C-2 and at terminal olefinic positions
Sarabia F and S'anchez-Ruiz A
The Journal of Organic Chemistry, 70(23), 9514-9520 (2005)
Total synthesis of bengamide E
Liu W, et al.
Tetrahedron Letters, 43(8), 1373-1375 (2002)
A diversity-oriented synthetic approach to bengamides
Sarabia F and S'anchez-Ruiz A
Tetrahedron Letters, 46(7), 1131-1135 (2005)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service