Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Palladium on alumina
Linear Formula:
Pd
Products
Brand
Sigma-Aldrich (7)
Sustainability
Feature
Melting Point (°C)
Physical Form
Quality Segment
Reaction Type
Reagent Type
Core
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
205710
Palladium on alumina,
extent of labeling: 5 wt. % loading, powder
Expand
View Pricing
205745
Palladium on alumina,
extent of labeling: 0.5 wt. % loading, 3.2 mm, pellets
Expand
View Pricing
908126
Palladium on alumina,
Evonik Noblyst
®
P1159 5% Pd
Expand
View Pricing
520675
Palladium on alumina,
extent of labeling: 0.5 wt. % loading, spheres
Expand
View Pricing
440086
Palladium on alumina,
extent of labeling: 10 wt. % loading, powder, reduced, dry
Expand
View Pricing
205702
Palladium on alumina,
extent of labeling: 1 wt. % loading, (Powder or beads)
Expand
View Pricing
761176
Palladium on alumina,
extent of labeling: 5 wt. % (dry basis), matrix activated alumina
Expand
View Pricing