Skip to Content
Merck
All Photos(3)

Key Documents

204374

Sigma-Aldrich

Silver acetate

99.99% trace metals basis

Synonym(s):

Acetic acid silver salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOAg
CAS Number:
Molecular Weight:
166.91
Beilstein:
3595636
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.99% trace metals basis

form

powder or crystals

reaction suitability

core: silver
reaction type: C-H Activation
reagent type: catalyst

SMILES string

CC(O[Ag])=O

InChI

1S/C2H4O2.Ag/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

CQLFBEKRDQMJLZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used in a novel preparation of highly reflective, conductive silvered polymer films. Also effectively catalyzes cycloaddition reactions of isocyanoacetates with a variety of olefins.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry of Materials, FL501-FL501 (1999)
Grigg, R. et al.
Tetrahedron, 55, 2025-2025 (1999)
Siddappa Patil et al.
Metallomics : integrated biometal science, 3(1), 74-88 (2010-12-08)
From the reaction of 1-methylimidazole (1a), 4,5-dichloro-1H-imidazole (1b(I)) and 1-methylbenzimidazole (1c) with p-cyanobenzyl bromide (2a), non-symmetrically substituted N-heterocyclic carbene (NHC) [(3a-c)] precursors, 5,6-dimethyl-1H-benzimidazole (1d) and 4,5-diphenyl-1H-imidazole (1e) with p-cyanobenzyl bromide (2a) and benzyl bromide (2b), symmetrically substituted N-heterocyclic carbene (NHC)
Tim Lancaster et al.
The Cochrane database of systematic reviews, 9(9), CD000191-CD000191 (2012-09-14)
Silver acetate produces an unpleasant taste when combined with cigarettes, thereby producing an aversive stimulus. It has been marketed in various forms with the aim of extinguishing the urge to smoke, by pairing the urge with an unpleasant stimulus. The
Axel Larena-Avellaneda et al.
Journal of vascular surgery, 50(4), 790-798 (2009-08-08)
Silver-coated vascular polyester prostheses were developed not only for the treatment of prosthetic graft infections, but also for use as prophylaxes. Although some studies describe the use of these prostheses in cases of infection, there are few data on their

Articles

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service