Recommended Products
Assay
99.95% trace metals basis
form
powder
impurities
≤550.0 ppm Trace Metal Analysis
density
3.11 g/mL at 25 °C (lit.)
SMILES string
[Rb+].[O-][N+]([O-])=O
InChI
1S/NO3.Rb/c2-1(3)4;/q-1;+1
InChI key
RTHYXYOJKHGZJT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Ox. Sol. 2
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Dalton transactions (Cambridge, England : 2003), 43(26), 10033-10039 (2014-05-29)
A new synthesis route for phosphates in an organic solvent at low temperatures is presented. The synthesis was done by dispersing a nitrate salt and phosphorus pentoxide in dimethyl sulfoxide. The synthesis can be performed under water-free conditions and yielded
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service