Skip to Content
Merck
All Photos(3)

Key Documents

203041

Sigma-Aldrich

Cesium nitrate

99.999% trace metals basis

Synonym(s):

Nitric acid cesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CsNO3
CAS Number:
Molecular Weight:
194.91
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
crystals and lumps

Quality Level

Assay

99.999% trace metals basis

form

crystals and lumps

impurities

≤15.0 ppm Trace Metal Analysis

mp

414 °C (lit.)

density

3.685 g/mL at 25 °C (lit.)

SMILES string

[Cs+].[O-]N(=O)=O

InChI

1S/Cs.NO3/c;2-1(3)4/q+1;-1

InChI key

NLSCHDZTHVNDCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cesium nitrate (CsNO3) is a class of perovskite based ferroelectric material that can be used as a highly efficient, nontoxic catalyst for the conversion of ketones and aldehydes to dihydroperoxides.

Application

CsNO3 can be used in a variety of lithium batteries such as lithium sulfur battery and lithium oxygen battery.
Used in the synthesis of cesium nitratocuprate, Cs2[Cu(NO3)4], which crystallizes in a square-planar coordination around the Cu center.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Ox. Sol. 1 - Repr. 2

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ferroelectric and phase transition studies in cesium nitrate: poly (vinyl alcohol) composite films
Nautiyal A, et al.
Applied Physics. A, Materials Science & Processing, 97(1), 205-210 (2009)
Cesium nitrate: As an efficient catalyst for synthesis of gem-dihydroperoxides from aldehydes and ketones using aqueous 30% H2O2
Khosravi K
Cogent Food & Agriculture, 1(1), 1002339-1002339 (2015)
A lithium-sulfur battery with a high areal energy density
Kim J, et al.
Advances in Functional Materials, 24(34), 5359-5367 (2014)
A molten salt lithium-oxygen battery
Giordani V, et al.
Journal of the American Chemical Society, 138(8), 2656-2663 (2016)
Morozov, I.V. et al.
Zeitschrift fur Anorganische und Allgemeine Chemie, 624, 1543-1543 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service