Skip to Content
Merck
All Photos(2)

Key Documents

203211

Sigma-Aldrich

Erbium(III) chloride hexahydrate

99.995% trace metals basis

Synonym(s):

Erbium trichloride hexahydrate, Erbium(3+) trichloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ErCl3·6H2O
CAS Number:
Molecular Weight:
381.71
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.995% trace metals basis
form:
solid

Quality Level

Assay

99.995% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: erbium

impurities

≤55.0 ppm Trace Rare Earth Analysis

SMILES string

O.O.O.O.O.O.Cl[Er](Cl)Cl

InChI

1S/3ClH.Er.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

UJBPGOAZQSYXNT-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Erbium (III) chloride hexahydrate is a pinkishcrystalline hydrate with unique magnetic and optical properties. It is soluble inwater. It is widely used in the fieldof optical fibers, luminescent devices, and organic synthesis. Its capacity for catalysis is enhanced by the dual Lewis acid and basebehavior.

Application

Erbium(III) chloride hexahydrate can be used:
  • As a chemical vapor deposition(CVD) precursor to fabricate erbium oxide thin films with high dielectric constant.
  • As a dopant to prepare specialty silica optical fibers.
  • As a catalyst for the rapid stereoselective synthesis of trans-4,5-diaminocyclopent-2-enones.
  • To fabricate a hybrid Erbium bis(pentafluorophenyl)phosphinate with long-lived infrared luminescence.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erbium(III) Chloride in Ethyl Lactate as a Smart Ecofriendly System for Efficient and Rapid Stereoselective Synthesis of trans-4,5-Diaminocyclopent-2-enones
Antonio Procopio, et al
ACS sustainable chemistry & engineering, 1, 541-544 (2013)
High-k erbium oxide film prepared by sol-gel method for low-voltage thin-film transistor
Guandong Wang, et al
Nanotechnology, 32, 215202-215202 (2021)
Multiple solution-doping in optical fibre fabrication II ? Rare-earth and aluminium co-doping
F.Z. Tang, et al.
Journal of Non-Crystalline Solids, 354, 1582-1590 (2008)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service