Skip to Content
Merck
All Photos(2)

Documents

198358

Sigma-Aldrich

Diethyl bis(hydroxymethyl)malonate

97%

Synonym(s):

Diethyl 2,2-bis(hydroxymethyl)malonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2)2C(CO2CH2CH3)2
CAS Number:
Molecular Weight:
220.22
Beilstein:
1788515
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

mp

49-51 °C (lit.)

SMILES string

CCOC(=O)C(CO)(CO)C(=O)OCC

InChI

1S/C9H16O6/c1-3-14-7(12)9(5-10,6-11)8(13)15-4-2/h10-11H,3-6H2,1-2H3

InChI key

WIOHBOKEUIHYIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for preparing 1,3-dioxanes from acetals, aldehydes, and ketones.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Chem. Soc. Perkin Trans. II, 1893-1893 (1991)
Stud. Univ. Babes-Bolyai, Chem., 33, 47-47 (1988)
Journal of the American Oil Chemists' Society, 70, 731-731 (1993)
Chem. Abstr., 111, 96357f-96357f (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service