Skip to Content
Merck
All Photos(1)

Key Documents

136441

Sigma-Aldrich

Dimethyl malonate

98%

Synonym(s):

Dimethyl propanedioate, Malonic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(COOCH3)2
CAS Number:
Molecular Weight:
132.11
Beilstein:
774261
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.413 (lit.)

bp

180-181 °C (lit.)

mp

−62 °C (lit.)

solubility

alcohol: miscible
diethyl ether: miscible
oil: miscible
water: slightly soluble

density

1.156 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

COC(=O)CC(=O)OC

InChI

1S/C5H8O4/c1-8-4(6)3-5(7)9-2/h3H2,1-2H3

InChI key

BEPAFCGSDWSTEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Dimethyl malonate undergoes enantioselective palladium-catalyzed allylic substitution reaction with 1,3-diphenylprop-2-enyl acetate to yield diastereomeric pure thienylpyridines.

Application

Dimethyl malonate was used in gold catalyzed oxidative esterification of glycerol, 1,2-propanediol and 1,3-propanediol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Chiral thienylpyridines as N-S ligands for asymmetric catalysis: Palladium-catalyzed allylic alkylation and copper-catalyzed cyclopropanation reactions.
Chelucci G, et al.
J. Mol. Catal. A: Chem., 197(1), 27-35 (2003)
Oxidation of glycerol and propanediols in methanol over heterogeneous gold catalysts.
Taarning E, et al.
Green Chemistry, 10(4), 408-414 (2008)
R Padmakumar et al.
Analytical biochemistry, 214(1), 318-320 (1993-10-01)
A rapid and high-yield synthesis of methylmalonyl coenzyme A is reported. The two-step procedure involves preparation of the thiophenyl ester of methylmalonic acid using dicyclohexylcarbodiimide as a condensing agent, followed by transesterification with coenzyme A, to yield methylmalonyl coenzyme A
Jeroen S Dickschat et al.
Chembiochem : a European journal of chemical biology, 5(6), 778-787 (2004-06-03)
The volatiles emitted from cell cultures of myxobacterium Myxococcus xanthus were collected by use of a closed-loop stripping apparatus (CLSA) and analyzed by GC-MS. Two new natural products, (S)-9-methyldecan-3-ol ((S)-1) and 9-methyldecan-3-one (2), were identified and synthesized, together with other
Catalytic oxidative decarboxylation of malic acid into dimethyl malonate in methanol with dioxygen.
Junxia Liu et al.
ChemSusChem, 5(11), 2151-2154 (2012-10-12)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service