197637
Imidazole sodium derivative
technical grade
Synonym(s):
Imidazolylsodium, Sodium imidazolide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C3H3N2Na
CAS Number:
Molecular Weight:
90.06
Beilstein:
3569312
EC Number:
MDL number:
UNSPSC Code:
12352005
eCl@ss:
39161001
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
technical grade
Quality Level
mp
284 °C (dec.) (lit.)
SMILES string
[Na]n1ccnc1
InChI
1S/C3H3N2.Na/c1-2-5-3-4-1;/h1-3H;/q-1;+1
InChI key
ITAWMPSVROAMOE-UHFFFAOYSA-N
Application
Imidazole sodium derivative (Imidazolylsodium) was used in the synthesis of arylazidoamorphigenin.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
F G Earley et al.
The Biochemical journal, 224(2), 525-534 (1984-12-01)
A photoaffinity-labelling analogue of the respiratory inhibitor rotenone was synthesized from the naturally occurring rotenoid amorphigenin. The analogue inhibits NADH-ubiquinone oxidoreductase activity at concentrations comparable with those of rotenone. Photolysis of the radiolabelled analogue bound to isolated NADH-ubiquinone oxidoreductase resulted
Sun Un
Inorganic chemistry, 52(7), 3803-3813 (2013-03-21)
A common feature of a large majority of the manganese metalloenzymes, as well as many synthetic biomimetic complexes, is the bonding between the manganese ion and imidazoles. This interaction was studied by examining the nature and structure of manganese(II) imidazole
Eleonora Petryayeva et al.
Langmuir : the ACS journal of surfaces and colloids, 29(3), 977-987 (2013-01-10)
Methods have been developed for the solid-phase detection of nucleic acids using mixed films of quantum dots (QDs) and oligonucleotide probes in microtiter plates. Polystyrene microwells were functionalized with multidentate imidazole ligands to immobilize QDs. Oligonucleotide hybridization was transduced using
Yun Dai et al.
Electrophoresis, 34(6), 833-840 (2013-01-22)
The first member of the single-isomer, dicationic cyclodextrin (CD) family, 6(A)-ammonium-6(C)-butylimidazolium-β-cyclodextrin chlorides (AMBIMCD), has been synthesized, analytically characterized, and used to separate a variety of acidic enantiomers and amino acids by CE. Starting from mono-6(A)-azido-β-cyclodextrin, the cationic imidazolium and ammonium
Lizhen Qiao et al.
Journal of chromatography. A, 1286, 137-145 (2013-03-16)
Hydrophilic interaction liquid chromatography (HILIC) has been widely used for separating polar compounds as a complement mode to reversed-phase liquid chromatography. The development of new stationary phases for HILIC is significant to improve the coverage of various polar and hydrophilic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service