Merck
  • Development and evaluation of new imidazolium-based zwitterionic stationary phases for hydrophilic interaction chromatography.
Development and evaluation of new imidazolium-based zwitterionic stationary phases for hydrophilic interaction chromatography.
Journal of chromatography. A (2013-03-16)
Lizhen Qiao, Abo Dou, Xianzhe Shi, Hua Li, Yuanhong Shan, Xin Lu, Guowang Xu
ABSTRACT
Hydrophilic interaction liquid chromatography (HILIC) has been widely used for separating polar compounds as a complement mode to reversed-phase liquid chromatography. The development of new stationary phases for HILIC is significant to improve the coverage of various polar and hydrophilic compounds. The present study described the preparation and application of novel imidazolium-based zwitterionic stationary phases. 1-Vinyl-3-(butyl-4-sulfonate) imidazolium was synthesized from 1-vinylimidazole and 1,4-butane sultone, then bonded to the surface of 3-mercaptopropyl modified silica particles (core-shell silica and totally porous silica) by "thiol-ene" click chemistry to obtain the resulting zwitterionic stationary phase with a positively charged imidazole ring and a negatively charged sulfonate group. The zwitterionic stationary phases exhibited good selectivity and favorable retention for a wide range of polar solutes (nucleosides, nucleic acid bases, benzoic organic acids, uric acid and its methyl derivatives, water-soluble vitamins) as compared to a bare silica column. The column efficiency could reach up to 100,000 theoretical plates/m with cytosine as the test solute. The retention changes of various types of test solutes were investigated under different chromatographic conditions including water content, pH, buffer salt concentration in mobile phase and column temperature. The results indicated that the retention of solutes on the stationary phase was the outcome of a mixed-mode retention mechanism (i.e. a combination of adsorptive and partitioning interactions). In conclusion, the new imidazolium-based zwitterionic stationary phases have shown excellent chromatographic behavior for a variety of polar solutes under HILIC mode, and have a great potential as a new type of stationary phases for HILIC.
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration), free-flowing, Redi-Dri
Imidazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole sodium derivative, technical grade
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole hydrochloride
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O