Recommended Products
Quality Level
Assay
97%
mp
264 °C (dec.) (lit.)
SMILES string
Cl[H].Cl[H].Nc1ccc(N)nc1
InChI
1S/C5H7N3.2ClH/c6-4-1-2-5(7)8-3-4;;/h1-3H,6H2,(H2,7,8);2*1H
InChI key
RWHNUWWUPZKDQP-UHFFFAOYSA-N
Application
2,5-Diaminopyridine dihydrochloride was used in the synthesis of:
- organic-inorganic hybrid compound, (C5H9N3)ZnCl4 .H2O
- (2-amino-5 ammoniopyridinium)[CuCl4]
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Three-Dimensional Network Polymeric Structure of (2-Amino-5 Ammoniopyridinium)[CuCl4]: Crystal Supramolecularity and Magnetic Properties.
Journal of Superconductivity and Novel Magnetism, 26(2), 437-442 (2013)
Preparation, crystal structure, and physical characterization of a new hybrid material (C5H9N3) ZnCl4? H2O.
Journal of Coordination Chemistry, 63(8), 1358-1368 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service