Skip to Content
Merck
All Photos(1)

Key Documents

D7148

Sigma-Aldrich

3,4-Diaminopyridine

≥98%

Synonym(s):

3,4-DAP, 4,5-Diaminopyridine, Amifampridine, DAP, Pyridine-4,5-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7N3
CAS Number:
Molecular Weight:
109.13
Beilstein:
110232
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Application

3,4-Diaminopyridine can be used as a heterocyclic building block to prepare:      
  • Imidazo[4,5-c]pyridine, an important scaffold used for the preparation of CP-885316.     
  • 9-Azajulolidine derivatives, which are used as efficient organocatalysts for acylation and aza-Morita-Baylis-Hillman reaction.      
  • 2-Phenylpyrido[3,4-b]pyrazine by condensation reaction with α-methylsulfinylacetophenone in benzene/acetic acid as a solvent.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regioselective Synthesis of an Imidazo [4, 5-c] pyridine through Selective Acylation of 3, 4-Diaminopyridine: Synthesis of CP-885,316
Caron S, et al.
Organic Process Research & Development, 10(2), 257-261 (2006)
Monika Cecilija Žužek et al.
Toxicology letters, 281, 95-101 (2017-09-25)
The effects of natural polymeric alkylpyridinium salt (nPoly-3-APS), a potent acetylcholinesterase inhibitor isolated from the marine sponge Reniera sarai, were studied on isolated mouse phrenic nerve-hemidiaphragm muscle preparations using electrophysiological approaches. nPoly-3-APS inhibited nerve-evoked isometric muscle twitch and tetanic contraction
Synthesis of 2-Arylpyrido [3, 4-b] pyrazine Derivatives through Condensation of 3, 4-Diaminopyridine with ?-Keto Sulfoxides
Kano S and Yuasa Y
Journal of Heterocyclic Chemistry, 20(3), 769-770 (1983)
Raman Tandon et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(20), 6435-6442 (2013-03-19)
New heterocyclic derivatives of 9-azajulolidine have been synthesized and characterized with respect to their nucleophilicity and Lewis basicity. The Lewis basicity of these bases as quantified through their theoretically calculated methyl-cation affinities correlate well with the experimentally measured reaction rates
Paul W Wirtz et al.
Expert review of clinical immunology, 6(6), 867-874 (2010-10-29)
The Lambert-Eaton myasthenic syndrome (LEMS) is an autoimmune disease in which antibodies against voltage-gated calcium channels inhibit cholinergic neurotransmission. LEMS is clinically characterized by muscle weakness and autonomic dysfunction. 3,4-diaminopyridine (3,4-DAP) blocks potassium channels in nerve terminals, resulting in an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service