Skip to Content
Merck
All Photos(1)

Key Documents

18509

Sigma-Aldrich

(+)-trans-Chrysanthemic acid

≥97.0% (GC)

Synonym(s):

(1R,3R)-trans-2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropane-1-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O2
CAS Number:
Molecular Weight:
168.23
Beilstein:
4904351
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

optical activity

[α]/D +14±1°, c = 2% in ethanol

refractive index

n20/D 1.477

SMILES string

C\C(C)=C/[C@@H]1[C@@H](C(O)=O)C1(C)C

InChI

1S/C10H16O2/c1-6(2)5-7-8(9(11)12)10(7,3)4/h5,7-8H,1-4H3,(H,11,12)/t7-,8+/m1/s1

InChI key

XLOPRKKSAJMMEW-SFYZADRCSA-N

General description

(+)-trans-Chrysanthemic acid is the acidic component of synthetic pyrethroid insecticides. It can be prepared from (+)-Δ3-carene.

Application

(+)-trans-Chrysanthemic acid may be used in the preparation of (+)-trans-pyrethric acid, a building block for rethrin II.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alain Krief et al.
Chemical communications (Cambridge, England), (39)(39), 4753-4755 (2008-10-03)
(1S)-cis-Chrysanthemic acid has been prepared in a few steps with complete control of the relative and absolute stereochemistry using regiocontrolled epoxide ring opening, diol mono-oxidation and cyclopropanation.
Gabriele Leng et al.
Toxicology letters, 162(2-3), 195-201 (2005-11-01)
Pyrethrum as well as synthetic pyrethroids like allethrin, resmethrin, phenothrin, tetramethrin, cyfluthrin, cypermethrin, deltamethrin or permethrin are among the insecticides most often used worldwide. With a sensitive and valid gas-chromatographic-high resolution mass spectrometric method, it is possible to detect all
Wolfgang Bicker et al.
Journal of chromatography. A, 1035(1), 37-46 (2004-05-01)
This study reports on the direct HPLC stereoisomer separation of selected pyrethroic acids employing commercial cinchona alkaloid derived chiral stationary phases (CSPs). cis/trans-Chrysanthemic acid (cis/trans-CA), cis/trans-chrysanthemum dicarboxylic acid (cis/trans-CDCA), cis/trans-permethrinic acid (cis/trans-PA), and fenvaleric acid (FA) were resolved into the
Mandar Paingankar et al.
Biotechnology letters, 27(23-24), 1909-1913 (2005-12-06)
Allethrin is a major mosquito repellent agent. To degrade allethrin present in used mats and the environment, a bacterium capable of utilizing allethrin was isolated. This isolate, an Acidomonas sp., grew in minimal medium with 16 mM: allethrin as sole
James R Durig et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 61(7), 1357-1373 (2005-04-12)
Infrared spectra of gaseous and solid 2-cyclopropylpropene (2-CPP, c-C3H5C (CH3)CH2) have been recorded from 3500 to 40 cm-1, and Raman spectra (3200-150 cm-1) of the liquid as well as mid-infrared spectra of 2-CPP in liquid krypton solution (from -105 to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service