Skip to Content
Merck
All Photos(2)

Documents

180343

Sigma-Aldrich

trans,trans-2,4-Hexadienal

95%

Synonym(s):

Sorbaldehyde, Sorbic aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=CHCHO
CAS Number:
Molecular Weight:
96.13
Beilstein:
1698401
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

95%

form

liquid

refractive index

n20/D 1.541 (lit.)

bp

69 °C/20 mmHg (lit.)

density

0.895 g/mL at 20 °C
0.871 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)\C=C\C=C\C

InChI

1S/C6H8O/c1-2-3-4-5-6-7/h2-6H,1H3/b3-2+,5-4+

InChI key

BATOPAZDIZEVQF-MQQKCMAXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans,trans-2,4-Hexadienal is an α,β-unsaturated aldehyde used as a food additive and as a starting material in various chemical and pharmaceutical industries.

Application

trans,trans-2,4-Hexadienal (sorbic aldehyde) was used in the synthesis of chiral cycloadducts.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

154.0 °F - closed cup

Flash Point(C)

67.77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

De novo asymmetric synthesis of two 5-amino-5, 6-dideoxy-D-allose derivatives.
Tetrahedron Letters, 35(31), 5653-5656 (1994)
Influence of 1-butanethiol and metal ions on hydrogenation of trans, trans-2, 4-hexadienal at platinum nanocatalysts
Du, YK and Xu, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 257, 75-78 (2005)
Atmospheric reactions between E, E-2, 4-hexadienal and OH, NO3 radicals and Cl atoms
Colmenar, et al.
Atmospheric Environment, 99, 159-167 (2014)
National Toxicology Program technical report series, (509)(509), 1-290 (2004-03-05)
2,4-Hexadienal, a colorless to yellow liquid with a pungent "green" or citrus odor, is used as a food additive for flavor enhancement, as a fragrance agent, as a starting material or intermediate in synthetic reactions in the chemical and pharmaceutical
Cell spreading on collagen that has been exposed to reactive aldehydes.
L Thorne et al.
Biochemical Society transactions, 20(4), 369S-369S (1992-11-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service