Skip to Content
Merck
All Photos(1)

Documents

177970

Sigma-Aldrich

Pyridine-d5

"100%", ≥99.96 atom % D

Synonym(s):

Pentadeuteropyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5D5N
CAS Number:
Molecular Weight:
84.13
Beilstein:
114377
EC Number:
MDL number:
UNSPSC Code:
12142201
eCl@ss:
39151701
PubChem Substance ID:
NACRES:
NA.11

Quality Level

isotopic purity

≥99.96 atom % D

Assay

≥99% (CP)

form

liquid

expl. lim.

0.34-6.3 % (lit.)

technique(s)

NMR: suitable

impurities

≤0.05% water
water

refractive index

n20/D 1.506 (lit.)

pH

8.5 (0.2 g/L)

bp

114.4 °C (lit.)

mp

-41 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

mass shift

M+5

SMILES string

[2H]c1nc([2H])c([2H])c([2H])c1[2H]

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H/i1D,2D,3D,4D,5D

InChI key

JUJWROOIHBZHMG-RALIUCGRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyridine-d5 is a deuterated form of pyridine having an isotopic purity of 99.96atom%D. It is 100% isotopically enriched NMR (Nuclear Magnetic Resonance) solvent. It is widely employed for high resolution NMR studies due to its high chemical and isotopic purity. The reaction between pyridine vapor and heavy water in the presence of a palladium catalyst has been reported to afford pyridine-d5. Infrared (IR), Raman (in the range 300-4000cm-1) and high-resolution (0.06cm-1) FT-IR (Fourier transform infrared) spectral studies of pyridine-d5 have been reported.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F

Flash Point(C)

17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vibrational spectra of C2v deuterium substituted pyridines. 4-pyridine-2, 3, 5, 6-d4 and pyridine-d5.
Stidham HD and Dilella DP.
Journal of Raman Spectroscopy, 9(4), 247-256 (1980)
The FT-IR spectra of pyridine and pyridine-d51.
Wong KN and Colson SD.
Journal of Molecular Spectroscopy, 104(1), 129-151 (1984)
Indra Prakash et al.
Natural product communications, 9(8), 1135-1138 (2014-09-23)
We report the isolation and complete structure of an isomer of rebaudioside D, known as rebaudioside D2. This novel steviol glycoside was isolated from a bioconversion reaction of rebaudioside A to rebaudioside D. Rebaudioside D2 possesses a relatively rare 1
Shao-Dan Chen et al.
Fitoterapia, 102, 67-73 (2015-02-24)
Five new diarylheptanoids (1-5), along with nine known ones (6-14), were isolated from the rhizomes of Curcuma kwangsiensis. Their structures were established on the basis of spectroscopic analyses. Compounds 1-3 were cyclic diarylheptanoids rarely discovered from C. kwangsiensis. Of all
The Vibrational Spectra of Pyridine and Pyridine-d 5.
Corrsin L, et al.
J. Chem. Phys. , 21(7), 1170-1176 (1953)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service