Skip to Content
Merck
All Photos(3)

Key Documents

166510

Sigma-Aldrich

1,1′-Diethyl-2,2′-carbocyanine iodide

97%

Synonym(s):

Pinacyanol iodide, Quinaldine blue

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H25IN2
CAS Number:
Molecular Weight:
480.38
Beilstein:
4117070
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

97%

form

powder

mp

295 °C (dec.) (lit.)

solubility

ethanol: 10 mg/mL

λmax

614 nm

ε (extinction coefficient)

≥180000 at 602-608 nm in ethanol
≥80000 at 560-566 nm in ethanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[I-].CCN1\C(=C\C=C\c2ccc3ccccc3[n+]2CC)C=Cc4ccccc14

InChI

1S/C25H25N2.HI/c1-3-26-22(18-16-20-10-5-7-14-24(20)26)12-9-13-23-19-17-21-11-6-8-15-25(21)27(23)4-2;/h5-19H,3-4H2,1-2H3;1H/q+1;/p-1

InChI key

QWYZFXLSWMXLDM-UHFFFAOYSA-M

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kausik Manna et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 74(5), 1268-1274 (2009-10-28)
Interaction of pinacyanol chloride (PIN) with pure and binary mixtures of cetyltrimethylammonium bromide (CTAB) and sodium deoxycholate (NaDC) was spectroscopically studied. Interaction of PIN with pure NaDC produced a blue shifted metachromatic band (at approximately 502 nm), which gradually shifted
R Nandini et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 75(1), 14-20 (2009-12-01)
The interactions of Acridine Orange with Sodium Alginate and Pinacyanol Chloride with Heparin have been investigated by spectrophotometric method. The polymers induce metachromasy in the dye as evidenced from the considerable blue shift in the absorption maxima of the corresponding
R K Narayan
Stain technology, 55(1), 9-11 (1980-01-01)
A technique is presented for rapid C-banding of eukaryotic chromosomes with pinacyanol chloride. This technique has given excellent definition and clarity to chromosome bands in plant and animal chromosomes. In permanent mounts the chromosomes did not fade after storage for
P G Krüger et al.
Acta neurologica Scandinavica, 81(1), 31-36 (1990-01-01)
In the brains of 7 patients with multiple sclerosis, mast cells were observed within the demyelinated plaques, in the border zone of the plaques as well as in seemingly normal white matter. The cells were mostly located in close connection
N Christensson et al.
The journal of physical chemistry. B, 115(18), 5383-5391 (2011-02-19)
In this work we analyze how nuclear coherences modulate diagonal and off-diagonal peaks in two-dimensional electronic spectroscopy. 2D electronic spectra of pinacyanol chloride are measured with 8 fs pulses, which allows coherent excitation of the 1300 cm(-1) vibrational mode. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service