Skip to Content
Merck
All Photos(2)

Key Documents

173754

Sigma-Aldrich

3,3′-Diethylthiadicarbocyanine iodide

Dye content 98 %

Synonym(s):

3-Ethyl-2-[5-(3-ethyl-2(3H)-benzothiazolylidene)-1,3-pentadienyl]benzothiazolium iodide, DTDCI, DiSC2(5)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H23IN2S2
CAS Number:
Molecular Weight:
518.48
Beilstein:
3838938
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

composition

Dye content, 98%

mp

249 °C (dec.) (lit.)

λmax

655 nm

SMILES string

[I-].CCN1/C(Sc2ccccc12)=C\C=C\C=C\c3sc4ccccc4[n+]3CC

InChI

1S/C23H23N2S2.HI/c1-3-24-18-12-8-10-14-20(18)26-22(24)16-6-5-7-17-23-25(4-2)19-13-9-11-15-21(19)27-23;/h5-17H,3-4H2,1-2H3;1H/q+1;/p-1

InChI key

MNQDKWZEUULFPX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

DTDCi is used as;
  • Fluorescent contrast agent in three-dimensional fluorescence lifetime tomography.
  • Dye for rapid genetic screening.

Features and Benefits

High quantum yield of photoisomerisation and low triplet quantum yield at direct photoexcitation. Highly sensitive fluorescent dye.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mengjia Xu et al.
Chembiochem : a European journal of chemical biology, 21(18), 2667-2675 (2020-04-19)
Peptide nucleic acids (PNAs), the synthetic DNA mimics that can bind to oligonucleotides to form duplexes, triplexes, and quadruplexes, could be advantageous as probes for nucleic acid sequences owing to their unique physicochemical and biochemical properties. We have found that
Photophysical processes of polymethine dyes. An absorption, emission, and optoacoustic study on 3, 3'-diethylthiadicarbocyanine iodide.
Bilmes GM, et al.
The Journal of Physical Chemistry, 93(18), 6696-6699 (1989)
Ryan M Williams et al.
Hypertension (Dallas, Tex. : 1979), 71(1), 87-94 (2017-11-15)
Direct targeting to the kidneys is a promising strategy to improve drug therapeutic index for the treatment of kidney diseases. We sought to investigate the renal selectivity and safety of kidney-targeted mesoscale nanoparticle technology. We found that direct intravenous administration
Jiang Pi et al.
Materials science & engineering. C, Materials for biological applications, 103, 109777-109777 (2019-07-28)
Tuberculosis (TB), caused by M.tuberculosis (Mtb), has become a top killer among infectious diseases. Enhancing the ability of anti-TB drugs to kill intracellular Mtb in host cells remains a big challenge. Here, an innovative nano-system was developed to increase drug
Complex formation between 3; 3'-diethylthiacarbocyanine iodide and DNA and its investigation in aqueous solution
Anikovsky, M. Y., Tatikolov, A. S., & Kuzmin, V. A.
International Journal of Photoenergy, 1(1), 35-39 (1999)

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service