Skip to Content
Merck
All Photos(1)

Key Documents

164216

Sigma-Aldrich

1-Phenylbiguanide

98%

Synonym(s):

1-(Diaminomethylidene)-2-phenylguanidine, 1-Carbamimidamido-N-phenylmethanimidamide, N-Phenyl-N′-guanylguanidine, N-Phenylbiguanide, N-Phenylimidodicarbonimidic diamide, N1-Phenylbiguanide, PBG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHC(=NH)NHC(=NH)NH2
CAS Number:
Molecular Weight:
177.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

135-142 °C (lit.)

solubility

alcohol: freely soluble
water: freely soluble

functional group

amine

SMILES string

NC(=N)\N=C(/N)Nc1ccccc1

InChI

1S/C8H11N5/c9-7(10)13-8(11)12-6-4-2-1-3-5-6/h1-5H,(H6,9,10,11,12,13)

InChI key

CUQCMXFWIMOWRP-UHFFFAOYSA-N

Gene Information

mouse ... Htr3a(15561)

General description

1-Phenylbiguanide is a 5-hydroxytryptamine receptor agonist and increases extracellular dopamine release in rat nucleus accumbens in vivo. It serves as building block for synthesis of heterocyclic compounds.

Application

1-Phenylbiguanide was used in one pot synthesis of :
  • 4-chloro-2,5-dihydro-2,5-dioxonaphtho[1,2-d]imidazole-3-carboxylic acid phenyl amide
  • 6-chloro-8-phenylamino-9H-7,9,11-triaza-cyclohepta[a]naphthalene-5,10-dione
  • 4-dimethylamino-5,10-dioxo-2-phenylimino-5,10-dihydro-2H-benzo[g]quinazoline-1-carboxylic acid amide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction of 2, 3-Dichloro-1, 4-Naphthoquinone with 1-Phenylbiguanide and 2-Guanidinebenzimidazole.
El-Shaieb KM.
J. Chin. Chem. Soc., 54(5), 1353-1358 (2007)
Irena Matulková et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 170, 256-266 (2016-07-29)
Three polymorphic modifications of bis(N-phenylbiguanidium(1+)) oxalate are reported, and their characterization is discussed in this paper. The non-centrosymmetric bis(N-phenylbiguanidium(1+)) oxalate (I), which was obtained from an aqueous solution at 313K, belongs to the monoclinic space group Cc (a=6.2560(2)Å, b=18.6920(3)Å, c=18.2980(5)Å
J P Chen et al.
Brain research, 543(2), 354-357 (1991-03-15)
The serotonin-3 (5-HT3) agonist 1-phenylbiguanide (0.1-1.0 mM in perfusate) caused a robust, dose-dependent enhancement of extracellular dopamine content in nucleus accumbens as measured by in vivo microdialysis. This action was antagonized by co-perfusion of the 5-HT3 antagonists zacopride and GR38032F
Debra I Diz et al.
Journal of cardiovascular pharmacology, 51(6), 542-548 (2008-05-14)
Endogenous angiotensin (Ang)-(1-7) enhances, while Ang II attenuates, baroreceptor sensitivity (BRS) for reflex control of heart rate (HR) in Sprague-Dawley (SD) rats. In (mRen2)27 renin transgenic rats [(mRen2)], there is overexpression of the mouse Ren2 gene in brain, leading to
Florence Netzer et al.
The European journal of neuroscience, 29(10), 2017-2028 (2009-05-21)
Stimulation of the dorsolateral periaqueductal gray matter (dlPAG) and the B3 cell group inhibits the cardiovagal component of the baroreflex in rats. Our aim was to determine whether the defence reaction induces similar modulatory effects on the cardiac response of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service