Skip to Content
Merck
All Photos(1)

Key Documents

153095

Sigma-Aldrich

4-Methylcyclohexanol, mixture of cis and trans

98%

Synonym(s):

Hexahydro-p-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H10OH
CAS Number:
Molecular Weight:
114.19
Beilstein:
1445998
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.94 (vs air)

Quality Level

vapor pressure

1.5 mmHg ( 30 °C)

Assay

98%

autoignition temp.

563 °F

refractive index

n20/D 1.458 (lit.)

bp

171-173 °C (lit.)

density

0.914 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC1CCC(O)CC1

InChI

1S/C7H14O/c1-6-2-4-7(8)5-3-6/h6-8H,2-5H2,1H3

InChI key

MQWCXKGKQLNYQG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methylcyclohexanol, mixture of cis and trans was used to study the effects of 4-Methylcyclohexanol on oviposition.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Felipe Delestro et al.
Scientific reports, 10(1), 7153-7153 (2020-04-30)
How does the concerted activity of neuronal populations shape behavior? Impediments to address this question are primarily due to critical experimental barriers. An integrated perspective on large scale neural information processing requires an in vivo approach that can combine the
Studies of 4-methylcyclohexanol: an Aedes triseriatus (Diptera: Culicidae) oviposition attractant.
M D Bentley et al.
Journal of medical entomology, 19(5), 589-592 (1982-10-14)
Jianzeng Guo et al.
Science (New York, N.Y.), 309(5732), 307-310 (2005-07-09)
Different modalities of sensation interact in a synergistic or antagonistic manner during sensory perception, but whether there is also interaction during memory acquisition is largely unknown. In Drosophila reinforcement learning, we found that conditioning with concurrent visual and olfactory cues
J R Linley
Medical and veterinary entomology, 3(4), 347-352 (1989-10-01)
Laboratory experiments tested the effects of p-cresol or 4-methylcyclohexanol at concentrations of 1, 10 and 50 ppm, on oviposition by the mosquitoes Toxorhynchites brevipalpis Theobald, Tx. amboinensis (Doleschall) and Tx. splendens (Wiedemann). A 5 + 5 ppm mixture of the
M Kutsukake et al.
Gene, 245(1), 31-42 (2000-03-14)
We characterized molecular profiles of a new olfactory mutant line, honoka (hono), which was found among 500 viable P-element insertion lines screened first by 5-bromo-4-chloro-3-indrolyl-beta-D-galactopyranoside (X-gal) staining on the third segment of the antenna, and then by behavioral assays to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service