Skip to Content
Merck

Skip To

1613757

USP

Sodium carbonate anhydrous

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Sodium carbonate, Calcined soda, Carbonic acid disodium salt, Soda ash

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
4154566
MDL number:
eCl@ss:
38020205
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

sodium carbonate

manufacturer/tradename

USP

mp

851 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C(O[Na])O[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sodium carbonate anhydrous USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Sodium Nitrate

Biochem/physiol Actions

Buffer component, may be used for the removal of peripheral membrane proteins.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.063981.0240513418-M
description

United States Pharmacopeia (USP) Reference Standard

description

anhydrous, EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JP, NF

description

volumetric standard, secondary reference material for acidimetry, traceable to NIST SRM Certipur®

description

puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

-

grade

-

grade

-

format

neat

format

-

format

-

format

-

application(s)

pharmaceutical (small molecule)

application(s)

liquid formulation
pharmaceutical
solid formulation

application(s)

-

application(s)

-

mp

851 °C (lit.)

mp

851 °C (lit.)

mp

851 °C (lit.)

mp

851 °C (lit.)

storage temp.

2-8°C

storage temp.

2-25°C

storage temp.

15-25°C

storage temp.

-


Still not finding the right product?

Explore all of our products under Sodium carbonate anhydrous


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters