Skip to Content
Merck
All Photos(1)

Key Documents

151955

Sigma-Aldrich

Nitrobenzene-d5

99.5 atom % D

Synonym(s):

Pentadeuteronitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D5NO2
CAS Number:
Molecular Weight:
128.14
Beilstein:
1455693
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99.5 atom % D

Quality Level

Assay

99% (CP)

form

liquid

expl. lim.

0.34-6.3 % (lit.)

technique(s)

NMR: suitable

impurities

≤0.5% water
water

refractive index

n20/D 1.5498 (lit.)

bp

88 °C/12 mmHg (lit.)

mp

6 °C (lit.)

density

1.253 g/mL at 25 °C (lit.)

mass shift

M+5

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])[N+]([O-])=O

InChI

1S/C6H5NO2/c8-7(9)6-4-2-1-3-5-6/h1-5H/i1D,2D,3D,4D,5D

InChI key

LQNUZADURLCDLV-RALIUCGRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitrobenzene-d5 is also referred to as pentadeuteronitrobenzene. Structure of the complexes between tetramethyl p-tert-butylcalix[4]arene tetraketone and hydroxonium cation H3O+ in nitrobenzene-d5 have been investigated by NMR (Nuclear Magnetic Resonance) spectroscopy and DFT (Density Functional Theory) calculations. It participates in the preparation of 2- and 3-phenyl-d5-thiophenes.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Repr. 1B - STOT RE 1 Inhalation

Target Organs

Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

190.4 °F

Flash Point(C)

88 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protonated tetramethyl p-tert-butylcalix [4] arene tetraketone: NMR evidence and probable structures.
Kriz J, et al.
Monatshefte fur Chemie / Chemical Monthly, 138(8), 735-740 (2007)
Vibrational spectra of nitrobenzene-d5.
Laposa JD.
Spectrochimica Acta Part A: Molecular Spectroscopy, 35(1), 65-71 (1979)
Arylation by aromatic nitro compounds at high temperatures. VII. Reactions of nitrobenzene and nitrobenzene-d5 with thiophene.
Fields EK and Meyerson S.
The Journal of Organic Chemistry, 35(1), 67-70 (1970)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service