Skip to Content
Merck
All Photos(1)

Documents

150215

Sigma-Aldrich

9(10H)-Acridanone

99%

Synonym(s):

Acridone, 9,10-Dihydro-9-oxoacridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9NO
CAS Number:
Molecular Weight:
195.22
Beilstein:
7104
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

>300 °C (lit.)

λmax

380 nm
399 nm (2nd)

SMILES string

O=C1c2ccccc2Nc3ccccc13

InChI

1S/C13H9NO/c15-13-9-5-1-3-7-11(9)14-12-8-4-2-6-10(12)13/h1-8H,(H,14,15)

InChI key

FZEYVTFCMJSGMP-UHFFFAOYSA-N

Gene Information

human ... ABCB1(5243)

Looking for similar products? Visit Product Comparison Guide

Application

9(10H)-Acridanone (acridone) was used in the preparation of methyl 9,10-dihydro-9-oxoacridine-10-pentanoate.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacob M Goldberg et al.
Journal of the American Chemical Society, 134(14), 6088-6091 (2012-04-05)
Fluorescent probe pairs that can be selectively excited in the presence of Trp and Tyr are of great utility in studying conformational changes in proteins. However, the size of these probe pairs can restrict their incorporation to small portions of
Vishal P Zambre et al.
Journal of molecular graphics & modelling, 29(2), 229-239 (2010-08-10)
G-quadruplex structures of DNA represent a potentially useful target for anticancer drugs. Telomerase enzyme, involved in immortalization of cancer cells is inhibited by stabilization of G-quadruplex at the ends of chromosomes. Anthraquinone and acridone derivatives are promising G-quadruplex ligands as
Alessandra Di Veroli et al.
Environmental pollution (Barking, Essex : 1987), 166, 212-217 (2012-04-21)
The aim of the present study was to improve the cause-effect relationship between toxicant exposure and chironomid mouthpart deformities, by linking induction of mouthpart deformities to contaminated field sediments, metal mixtures and a mutagenic polycyclic aromatic compound metabolite (acridone). Mouthpart
Jatinder Kaur et al.
Chemical communications (Cambridge, England), 47(15), 4472-4474 (2011-03-08)
Acridones carrying an appropriate substituent at N-10 showed significant fluorescence changes on interacting with ATP in HEPES buffer at pH 7.2. The selectivity and sufficient binding of these probes with ATP could be useful for monitoring of metabolic processes.
Anton V Dubrovskiy et al.
The Journal of organic chemistry, 77(24), 11232-11256 (2012-12-05)
A novel, efficient route to biologically and pharmaceutically important o-(dimethylamino)aryl ketones, acridones, acridinium salts, and 1H-indazoles has been developed starting from readily available hydrazones of aldehydes and o-(trimethylsilyl)aryl triflates. The reaction proceeds through arynes under mild conditions, tolerates a wide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service