Skip to Content
MilliporeSigma

Skip To

E45708

1-Ethylpiperidine

99%

Synonym(s):

N-Ethylpiperidine, NSC 2090

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C7H15N
CAS Number:
Molecular Weight:
113.20
UNSPSC Code:
12352100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
212-161-6
Beilstein/REAXYS Number:
102643
MDL number:
Assay:
99%
Form:
liquid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

99%

form

liquid

refractive index

n20/D 1.444 (lit.)

bp

131 °C (lit.)

density

0.824 g/mL at 25 °C (lit.)

SMILES string

CCN1CCCCC1

InChI

1S/C7H15N/c1-2-8-6-4-3-5-7-8/h2-7H2,1H3

InChI key

HTLZVHNRZJPSMI-UHFFFAOYSA-N

Application

Reactant for:
Synthesis of multiprotected kanosamine
Selective acylation in peptide synthesis[1]
Reagent for:
Stereoselective aldol condensation reactions for synthesis of β-lactam antibiotics
Diastereoselective synthesis of aldols
Crossed Claisen ester condensation


signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

62.6 °F

flash_point_c

17 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters