Skip to Content
Merck
All Photos(4)

Documents

149837

Sigma-Aldrich

2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline

≥99%, for peptide synthesis

Synonym(s):

N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, EEDQ, Ethyl 1,2-dihydro-2-ethoxyquinoline-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17NO3
CAS Number:
Molecular Weight:
247.29
Beilstein:
533048
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

product name

2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, ≥99%

Assay

≥99%

form

solid

reaction suitability

reaction type: Coupling Reactions

mp

62-67 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CCOC1C=Cc2ccccc2N1C(=O)OCC

InChI

1S/C14H17NO3/c1-3-17-13-10-9-11-7-5-6-8-12(11)15(13)14(16)18-4-2/h5-10,13H,3-4H2,1-2H3

InChI key

GKQLYSROISKDLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline(EEDQ) is a stable, and readily available reagent used to prepare amides and peptides in high yields. At ambient temperature, EEDQ is soluble in an aqueous medium containing organic solvents but insoluble in water. It is used as a coupling agent in peptide synthesis.

Application

2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline is used as a coupling agent:
  • In the regioselective quaternization of chitosan and its amphiphilic derivatives.
  • In the preparation of amide-type S-MA derivative-modified QCM sensors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eur. J. Pharmacol., Molec. Pharmacol. Sec., 269, 25-25 (1994)
Analía Bortolozzi et al.
The international journal of neuropsychopharmacology, 13(10), 1299-1314 (2010-02-18)
Atypical antipsychotic drugs (APDs) increase dopamine (DA) release in prefrontal cortex (PFC), an effect probably mediated by the direct or indirect activation of the 5-HT(1A) receptor (5-HT(1A)R). Given the very low in-vitro affinity of most APDs for 5-HT(1A)Rs and the
L Carbonell et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 14(6), 497-502 (2004-12-14)
One of the most effective psychotherapeutic agents in the treatment of bipolar disease is lithium. Chronic lithium treatment affects some signal transduction mechanisms such as cAMP, cGMP, inositol 1,4,5 P(3), Gi protein, protein kinase C and can also modify gene
George Vaniotis et al.
Journal of molecular and cellular cardiology, 62, 58-68 (2013-05-21)
At the cell surface, βARs and endothelin receptors can regulate nitric oxide (NO) production. β-adrenergic receptors (βARs) and type B endothelin receptors (ETB) are present in cardiac nuclear membranes and regulate transcription. The present study investigated the role of the
New approach to the quaternization of chitosan and its amphiphilic derivatives
Stepnova EA, et al.
European Polymer Journal, 43(6), 2414-2421 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service