129976
2-Methyl-3-pentanol
≥99%
Synonym(s):
Ethyl isopropyl carbinol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CH2CH(OH)CH(CH3)2
CAS Number:
Molecular Weight:
102.17
Beilstein:
1718952
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥99%
form
liquid
refractive index
n20/D 1.417 (lit.)
bp
128 °C (lit.)
density
0.819 g/mL at 25 °C (lit.)
functional group
hydroxyl
SMILES string
CCC(O)C(C)C
InChI
1S/C6H14O/c1-4-6(7)5(2)3/h5-7H,4H2,1-3H3
InChI key
ISTJMQSHILQAEC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
2-Methyl-3-pentanol (Ethyl isopropyl carbinol) was detected by comprehensive two-dimensional gas chromatography-time-of-flight mass spectrometry (GC×GC-ToFMS).
Application
2-Methyl-3-pentanol (Ethyl isopropyl carbinol) was used as a volatile marker.
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
114.8 °F - closed cup
Flash Point(C)
46 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Angelo C Salvador et al.
PloS one, 8(4), e59338-e59338 (2013-04-25)
A novel approach based on headspace solid-phase microextraction (HS-SPME) combined with comprehensive two-dimensional gas chromatography-time-of-flight mass spectrometry (GC×GC-ToFMS) was developed for the simultaneous screening of microbial and mite contamination level in cereals and coffee beans. The proposed approach emerges as
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service