111600
2-Methyl-2-hexanol
97%
Synonym(s):
1,1-Dimethyl-1-pentanol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)3C(CH3)2OH
CAS Number:
Molecular Weight:
116.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
refractive index
n20/D 1.417 (lit.)
bp
141-142 °C (lit.)
density
0.812 g/mL at 25 °C (lit.)
functional group
hydroxyl
SMILES string
CCCCC(C)(C)O
InChI
1S/C7H16O/c1-4-5-6-7(2,3)8/h8H,4-6H2,1-3H3
InChI key
KRIMXCDMVRMCTC-UHFFFAOYSA-N
Related Categories
Application
2-Methyl-2-hexanol was used in the preparation of esterified phenol. It is used to study the effect of organic solvent on the equillibrium of lipase-catalyzed esterification of glycerol and decanoic acid.
2-Methyl-2-hexanol was used to study the mass accommodation coefficient (α) on 1-methylnaphthalene as a function of temperature (267 K to 298 K) for gas-phase of the product.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
105.8 °F - closed cup
Flash Point(C)
41 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A E Janseen et al.
Biotechnology and bioengineering, 41(1), 95-103 (1993-01-05)
The effect of organic solvents on the equilibrium position of lipase-catalyzed esterification of glycerol and decanoic acid has been investigated. The reaction is carried out in an aqueous-organic two-phase system. In polar solvents, high mole fractions of monoacylglycerol and low
An achiral, anticlinic-promoting, smectic liquid crystal architecture.
Thisayukta J and Samulski ET.
Journal of Materials Chemistry, 14(10), 1554-1559 (2004)
H Z Zhang et al.
The journal of physical chemistry. A, 109(17), 3941-3949 (2006-07-13)
Organic compounds are a significant component of tropospheric aerosols. In the present study, 1-methylnaphthalene was selected as a surrogate for aromatic hydrocarbons (PAHs) found in tropospheric aerosols. Mass accommodation coefficients (alpha) on 1-methylnaphthalene were determined as a function of temperature
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service