Skip to Content
Merck
All Photos(2)

Key Documents

112976

Sigma-Aldrich

2,5-Di-tert-butylhydroquinone

99%

Synonym(s):

1,4-Dihydroxy-2,5-di-tert-butylbenzene, 2,5-Bis(1,1-dimethylethyl)-1,4-benzenediol, 2,5-Di-tert-butyl-1,4-benzenediol, 2,5-Di-tert-butyl-1,4-benzohydroquinone, 2,5-Di-tert-butyl-1,4-dihydroxybenzene, 2,5-Di-tert-butylquinol, 2,5-Dihydroxy-1,4-di-tert-butylbenzene, 2,5-Ditert-butylbenzene-1,4-diol, 3,6-Di-tert-butylhydroquinone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H2-1,4-(OH)2
CAS Number:
Molecular Weight:
222.32
Beilstein:
2049542
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

autoignition temp.

790 °F

mp

216-218 °C (lit.)

SMILES string

CC(C)(C)c1cc(O)c(cc1O)C(C)(C)C

InChI

1S/C14H22O2/c1-13(2,3)9-7-12(16)10(8-11(9)15)14(4,5)6/h7-8,15-16H,1-6H3

InChI key

JZODKRWQWUWGCD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,5-Di-tert-butylhydroquinone is the oxidation substrate used to measure the catalytic activity of the copper(II) enzyme-like catalysts.

Biochem/physiol Actions

2,5-Di-tert-butylhydroquinone specifically inhibits the sarcoplasmic reticulum (SR) Ca2+ uptake in the rat ventricle.
Blocks calcium-dependent insulin signaling in human beta cells

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salvatore Nesci et al.
Biochimie, 140, 73-81 (2017-06-28)
The mitochondrial F-ATPase can be activated either by the classical cofactor Mg
Weiwei Zhang et al.
Acta biomaterialia, 8(2), 897-903 (2011-10-11)
To improve the rates of both cupric ion release and the utilization of copper in non-porous copper/low-density polyethylene (Cu/LDPE) composite, a porous Cu/LDPE composite is proposed and developed in the present work. Here 2,5-di-tert-butylhydroquinone was chosen as the porogen, ethyl
W A Macdonald et al.
The Journal of physiology, 573(Pt 1), 187-198 (2006-03-25)
Slow-twitch mechanically skinned fibres from rat soleus muscle were bathed in solutions mimicking the myoplasmic environment but containing different [ADP] (0.1 microm to 1.0 mm). The effect of ADP on sarcoplasmic reticulum (SR) Ca2+-content was determined from the magnitude of
Joel Castro et al.
The Biochemical journal, 418(3), 553-566 (2008-11-15)
The question of whether the activation of SOCs (store-operated Ca(2+) channels) requires the whole or part of the ER (endoplasmic reticulum) has not been fully resolved. The role of a putative sub-compartment of the ER in SOC activation in liver
Francesco Tadini-Buoninsegni et al.
Molecular pharmacology, 73(4), 1134-1140 (2008-01-24)
The inhibitory effects of thapsigargin, cyclopiazonic acid, and 2,5-di(tert-butyl)hydroquinone, and 1,3-dibromo-2,4,6-tri(methylisothiouronium)benzene on the Ca(2+) ATPase were characterized by comparative measurements of sequential reactions of the catalytic and transport cycle, including biochemical measurements and detection of charge movements within a single

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service