All Photos(1)
About This Item
Linear Formula:
C6H5C(=NH)OCH2CH3 · HCl
CAS Number:
Molecular Weight:
185.65
Beilstein:
3913195
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥97.0% (AT)
mp
~125 °C (dec.)
functional group
ether
phenyl
SMILES string
Cl[H].CCOC(=N)c1ccccc1
InChI
1S/C9H11NO.ClH/c1-2-11-9(10)8-6-4-3-5-7-8;/h3-7,10H,2H2,1H3;1H
InChI key
MODZVIMSNXSQIH-UHFFFAOYSA-N
Related Categories
General description
Ethyl benzimidate hydrochloride reacts with (R)-ethyl cysteine hydrochloride in ethanol to yield (4R)-ethyl 2-phenyl-4,5-dihydrothiazole-4-carboxylate. It reacts with D-Penicillamine methyl ester hydrochloride and triethylamine to yield methyl-5,5- dimethyl-2-phenyl-2-thiazoline-4-carboxylate.
Application
Intermediate for synthesis
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Synthesis of Substituted Penicillins and Simpler Structural Analogs. III. Phthalimido ?-Lactam-Thiazolidines Derived from Penicillamine.
Sheehan JC, et al.
Journal of the American Chemical Society, 73(9), 4373-4375 (1951)
Satendra Singh et al.
The Journal of organic chemistry, 69(13), 4551-4554 (2004-06-19)
(1R)-(+)-2,10- and (1S)-(-)-2,10-camphorsultam were acylated with ethyl 2-phenylthiazoline 4-carboxylate to afford (+)- and (-)-2-phenylthiazolinylcamphorsultam, which were stereoselectively alkylated with MeI in the presence of n-BuLi. Alkylation of these phenylthiazolinylcamphorsultams occurred from the beta-face rather than alpha-face, resulting in the formation
Aerobic dissipation of the novel cyanoacrylate fungicide phenamacril in soil and sludge incubations.
Søren S Donau et al.
Chemosphere, 233, 873-878 (2019-07-26)
The cyanoacrylate ethyl (2Z)-3-amino-2-cyano-3-phenylacrylate (phenamacril), has been introduced as an effective agent against several fungi species belonging to the Fusarium genus. However, in current literature, knowledge about the environmental behavior of this fungicide is limited and there are no data
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service