Skip to Content

8.10647

Kryptofix® 222

for synthesis

Synonym(s):

Kryptofix® 222, 4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H36N2O6
CAS Number:
Molecular Weight:
376.49
UNSPSC Code:
12352005
EC Index Number:
245-962-4
NACRES:
NA.22
MDL number:
Assay:
99%
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

99%

form

powder

potency

300-2000 mg/kg LD50, oral (Rat)

reaction suitability

core: cryptand

mp

70-73 °C

storage temp.

2-8°C

SMILES string

N21CCOCCOCCN(CCOCCOCC2)CCOCCOCC1

InChI

1S/C18H36N2O6/c1-7-21-13-14-24-10-4-20-5-11-25-17-15-22-8-2-19(1)3-9-23-16-18-26-12-6-20/h1-18H2

InChI key

AUFVJZSDSXXFOI-UHFFFAOYSA-N

Application

Kryptofix® 222 for synthesis. CAS 23978-09-8, molar mass 376.49 g/mol.

Analysis Note

Purity (GC): ≥ 99
Identity (IR-spectrum): passes test
Melting range: 70 - 73

Legal Information

Kryptofix is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Kryptofix® 222


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library