Skip to Content
Merck
All Photos(3)

Documents

114618

Sigma-Aldrich

Isatin

97%

Synonym(s):

2,3-Diketoindoline, 2,3-Dioxo-2,3-dihydroindole, 2,3-Dioxoindoline, o-Aminobenzoylformic anhydride, Isatic acid lactam, Isatinic acid anhydride, 2,3-Indolinedione, Isatine, NSC 9262

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5NO2
CAS Number:
Molecular Weight:
147.13
Beilstein:
383659
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

193-195 °C (dec.) (lit.)

solubility

boiling water: soluble
diethyl ether: soluble
soluble

SMILES string

O=C1Nc2ccccc2C1=O

InChI

1S/C8H5NO2/c10-7-5-3-1-2-4-6(5)9-8(7)11/h1-4H,(H,9,10,11)

InChI key

JXDYKVIHCLTXOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Isatin can be used as a reactant to prepare:
  • Phthalazinone derivatives.
  • Spirooxindole derivatives.
  • Agents against multidrug-resistant cells expressing P-glycoprotein.
  • Thiazolidinones spiro-fused to indolin-2-ones as potent and selective inhibitors of Mycobacterium tuberculosis protein tyrosine phosphatase B.
  • Triazole-isatine compounds as potential antibacterial and antifungal agents.
  • Biologically relevant scaffolds such as spiro[indole-thiazolidinones].

It can be a reactant for:
  • Cascade reactions with heterocyclic ketene aminals.
  • Knoevenagel condensation reactions.

Isatin can also be used as a chromatographic spray reagent for amino acids, and also as a reference material in the preparation of indigo and Maya Blue. It is responsible for the yellow color exhibited by the "Maya Yellow" pigment.
Isatin undergoes Pd(II)-catalyzed C-H addition by direct sp2/sp3 C-H bond activation to form 3-substituted-3-hydroxy-2-oxindoles. Isatin derivatives (bis-Schiff base) are synthesized by the condensation of the natural or synthetic isatins with hydrazine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Novel Synthesis of Some Phthalazinone Derivatives
Mahmoud, M. R.; et al.
Chin. J. Chem., 29, 1446-1450 (2011)
Fuchao Yu et al.
Organic letters, 13(18), 4782-4785 (2011-08-19)
A concise and efficient route for the synthesis of highly substituted imidazopyrroloquinoline derivatives by simply refluxing a reaction mixture of different types of isatins and heterocyclic ketene aminals (HKAs) by acetic acid was developed. This method is suitable for combinatorial
Ozil, M.; et al.
J. Chem. Res. (M), 35, 268-268 (2011)
Gang-Wei Wang et al.
Organic letters, 15(20), 5270-5273 (2013-10-05)
The first Pd(II)-catalyzed C-H addition to isatins by direct sp(2)/sp(3) C-H bond activation for the construction of 3-substituted-3-hydroxy-2-oxindoles is reported. The bidentate nitrogen ligands were found to promote this reaction. Specifically, the preliminary bioassay indicated that 3-(5-chlorobenzoxazole)-3-hydroxy-N-benzyl-2-oxindole (2w) is a
Versatile three component procedure for combinatorial synthesis of biologically relevant scaffold spiro[indole-thiazolidinones] under aqueous conditions
Dandia, A.; et al.
Green Chemistry, 13, 1852-1852 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service