Skip to Content
Merck
All Photos(1)

Documents

112062

Sigma-Aldrich

N-Ethyldiethanolamine

98%

Synonym(s):

2,2′-Ethyliminodiethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2CH2)2NC2H5
CAS Number:
Molecular Weight:
133.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.4665 (lit.)

bp

246-252 °C (lit.)

mp

−50 °C (lit.)

density

1.014 g/mL at 25 °C (lit.)

SMILES string

CCN(CCO)CCO

InChI

1S/C6H15NO2/c1-2-7(3-5-8)4-6-9/h8-9H,2-6H2,1H3

InChI key

AKNUHUCEWALCOI-UHFFFAOYSA-N

General description

N-Ethyldiethanolamine can be identified in water, urine and blood samples by gas chromatography-mass spectrometry (GC-MS).
N-Ethyldiethanolamine is a precursor of VX type nerve agent.

Application

N-Ethyldiethanolamine was used in the analysis of urine for assaying nitrogen mustard metabolites.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

255.2 °F - closed cup

Flash Point(C)

124 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prabhat Garg et al.
Journal of chromatography. A, 1216(45), 7906-7914 (2009-10-03)
N,N-Dialkylamino alcohols, N-methyldiethanolamine, N-ethyldiethanolamine and triethanolamine are the precursors of VX type nerve agents and three different nitrogen mustards respectively. Their detection and identification is of paramount importance for verification analysis of chemical weapons convention. GC-FTIR is used as complimentary
Buddhadeb Chandra et al.
Rapid communications in mass spectrometry : RCM, 34(3), e8586-e8586 (2019-09-12)
Nitrogen mustards (NMs) are vesicant class of chemical warfare agents. From the viewpoint of the Chemical Weapons Convention partially hydrolyzed products of nitrogen mustards (pHpNMs) are considered as important markers of nitrogen mustard exposure. The detection of pHpNMs from biological
Buddhadeb Chandra et al.
Analytical methods : advancing methods and applications, 12(36), 4447-4456 (2020-08-29)
The development and optimization of an analytical method for the detection and identification of reactive metabolite of organochlorine chemical warfare agent nitrogen mustards (NMs), 2-[(2-chloroethyl)(alkyl)amino]ethanol (CEAAE), known as half nitrogen mustard, in blood samples is presented, herein. In this study
Buddhadeb Chandra et al.
Rapid communications in mass spectrometry : RCM, 34(12), e8777-e8777 (2020-03-08)
Analytical methods for the detection and identification of half nitrogen mustards (halfNMs), i.e., partially hydrolyzed products of nitrogen mustards (pHpNMs), using silyl derivatives are often associated with low sensitivity and selectivity. In order to overcome these limitations, the derivatization of
Shanker Karunanithi et al.
eNeuro, 7(1) (2020-02-06)
Propofol is the most common general anesthetic used for surgery in humans, yet its complete mechanism of action remains elusive. In addition to potentiating inhibitory synapses in the brain, propofol also impairs excitatory neurotransmission. We use electrophysiological recordings from individual

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service