Merck
  • In vitro degradation behavior of Fe-20 Mn-1.2C alloy in three different pseudo-physiological solutions.
In vitro degradation behavior of Fe-20 Mn-1.2C alloy in three different pseudo-physiological solutions.
Materials science & engineering. C, Materials for biological applications (2016-02-04)
Essowè Mouzou, Carlo Paternoster, Ranna Tolouei, Agung Purnama, Pascale Chevallier, Dominique Dubé, Frédéric Prima, Diego Mantovani
ABSTRACT
High manganese austenitic steels such as Fe-20Mn-1.2C alloys are among the most promising candidates for biodegradable stents applications due to their high strength, high ductility and their chemical composition. In the current work, 14 day static in-vitro tests were performed in controlled atmosphere to assess the degradation behavior in three common pseudo-physiological solutions, i.e. commercial Hanks' (CH), modified Hanks' (MH) and albumin-enriched Dulbecco's modified phosphate buffered saline (DPBS) solutions. The degraded samples surfaces as well as the degradation products were characterized by X-ray diffraction (XRD), scanning electron microscopy (SEM) and Fourier transform infrared spectroscopy (FTIR). Degradation of material and degradation products are shown to be strongly dependent on the test medium due to the presence of different ionic species such as HCO3(-), CO3(2-), Cl(-), Ca(2+) or phosphate groups. In both MH and CH solutions, the increased content of HCO3(-) ions seems to promote MnCO3 crystal growth on sample surfaces whereas the presence of albumin and high content of phosphate ions promotes the formation of an amorphous layer rich in phosphates, iron and manganese.
Sigma-Aldrich
HEPES sodium salt, ≥99.0% (titration)
Sigma-Aldrich
Hanks′ Balanced Salts, Modified, without phenol red and sodium bicarbonate, powder, suitable for cell culture
SAFC
Hanks′ Balanced Salts, Modified, without calcium chloride, magnesium sulfate and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, powder, suitable for cell culture