Skip to Content
Merck
All Photos(5)

Documents

H7006

Sigma-Aldrich

HEPES sodium salt

≥99.0% (titration)

Synonym(s):

Sodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulfonate, 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17N2NaO4S
CAS Number:
Molecular Weight:
260.29
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99.0% (titration)

form

powder

storage condition

dry at room temperature

technique(s)

HPLC: suitable

color

white

pH

10-12 (1 M in water)

useful pH range

6.8-8.2

pKa (25 °C)

7.5

solubility

water: 0.1 M, clear, colorless

suitability

suitable for mass spectrometry

application(s)

diagnostic assay manufacturing
general analytical (
)
life science and biopharma
metabolomics

storage temp.

room temp

SMILES string

[Na+].OCCN1CCN(CC1)CCS([O-])(=O)=O

InChI

1S/C8H18N2O4S.Na/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14;/h11H,1-8H2,(H,12,13,14);/q;+1/p-1

InChI key

RDZTWEVXRGYCFV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

HEPES salt, or 4-(2-hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, is a zwitterionic biological buffer that is widely used in biological and biochemical research. It has a buffering range of pH 6.8 to 8.2, making it ideal for a variety of research applications. HEPES is also a popular choice for buffering cell culture media, offering advantages such as lower cell toxicity in comparison to alternatives like Tris and phosphate.

Widely utilized in cell biology and biochemical research, HEPES proves its versatility in protein purification and extraction protocols, owing to its stability across a broad pH range and minimal metal ion binding. Additionally, HEPES proves valuable in various biological and biochemical processes, including immunoprecipitation, cell lysis, and live cell imaging, making it a versatile tool in diverse research applications.

Application

HEPES buffer finds application in:
  • Most studies with metal ions
  • Substitute for Tris and phosphate in studies with metal ions
  • Grinding buffer in plant studies
  • Running buffer in gel electrophoresis
  • Buffer in electroporation studies
  • Bradford or bicinchoninic acid (BCA) assays
  • Extraction buffer to prevent damage of proteins in red blood cells

It has been used:
  • as a buffer component in the preparation of a low sucrose buffer
  • as a buffer component in a modified Hanks′ solution, a pseudo-physiological solutions

Features and Benefits

  • Tested to confirm low levels of heavy metal contamination, ensuring suitability for various applications
  • Highly soluble in water
  • Negligible metal ion binding
  • Less toxic to cells than other buffers such as Tris and phosphate
  • Stable in a wide pH range

Physical form

Lyophilized, spray dried, or crystallized material.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nishant Mehta et al.
Cell reports, 28(10), 2509-2516 (2019-09-05)
V-domain immunoglobulin (Ig) suppressor of T cell activation (VISTA) is an immune checkpoint protein that inhibits the T cell response against cancer. Similar to PD-1 and CTLA-4, a blockade of VISTA promotes tumor clearance by the immune system. Here, we report a
Camila Jordão Candido et al.
Nutrients, 12(1) (2019-12-22)
Consumption of omega-3 (n-3) polyunsaturated fatty acids (PUFA) is related to improvement in the inflammatory response associated with decreases in metabolic disorders of obesity, such as low-grade inflammation and hepatic steatosis. Linseed (Linum usitatissimum) oil is a primary source of
Essowè Mouzou et al.
Materials science & engineering. C, Materials for biological applications, 61, 564-573 (2016-02-04)
High manganese austenitic steels such as Fe-20Mn-1.2C alloys are among the most promising candidates for biodegradable stents applications due to their high strength, high ductility and their chemical composition. In the current work, 14 day static in-vitro tests were performed
Ekaterina Eremenko et al.
STAR protocols, 2(3), 100643-100643 (2021-07-27)
The organization of chromatin structure plays a crucial role in gene expression, DNA replication, and repair. Chromatin alterations influence gene expression, and modifications could be associated with genomic instability in the cells during aging or diseases. Here, we provide a
Luke Grundy et al.
American journal of physiology. Renal physiology, 318(2), F298-F314 (2019-12-04)
Interstitial cystitis/bladder pain syndrome (IC/BPS) is a common chronic pelvic disorder with sensory symptoms of urinary urgency, frequency, and pain, indicating a key role for hypersensitivity of bladder-innervating sensory neurons. The inflammatory mast cell mediator histamine has long been implicated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service