Merck
  • Simple and robust monitoring of ethanol fermentations by capillary electrophoresis.
Simple and robust monitoring of ethanol fermentations by capillary electrophoresis.
Biotechnology and applied biochemistry (2014-07-22)
James D Oliver, Adam T Sutton, Naama Karu, Michael Phillips, Julie Markham, Paul Peiris, Emily F Hilder, Patrice Castignolles
ABSTRACT
Free-solution capillary electrophoresis (CE), or capillary zone electrophoresis, with direct UV detection was used for the first time for the determination of mono- and disaccharides, sugar alcohols, and ethanol in fermentation broths. Sample preparation proved to be minimal: no derivatization or specific sample purification was needed. The CE conditions can be adapted to the type of fermentation by simply altering the background electrolyte (BGE). KOH (130 mM) or NaOH (130 mM) as the BGE led to the fastest analysis time when monitoring simple fermentations. A mixture of 65 mM NaOH and 65 mM LiOH led to a 19% improvement in resolution for a complex mixture of carbohydrates. Quantification of a simple carbohydrate fermentation by CE showed values in close agreement with that of high-performance anion exchange chromatography and high-performance liquid chromatography (HPLC) on a cation exchange resin. For complex fermentations, quantification of carbohydrates by HPLC and CE led to similar results, whereas CE requires an injection volume of only 10-20 nL. Analysis of an ethanol fermentation of hydrolyzed plant fiber demonstrated the robustness of the separation and detection of carbohydrates, as well as ethanol. Ethanol determination is achieved by coupling the CE method to pressure mobilization, using the same instrument and the same sample.
Sigma-Aldrich
L-(+)-Arabinose, 99%
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 16O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
L-(+)-Arabinose, 98%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
L-(+)-Arabinose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Xylitol, ≥99% (GC)
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Sigma-Aldrich
D-(−)-Fructose, 98.0-102.0% dry basis, meets USP testing specifications
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
Lithium-7Li hydroxide monohydrate, 99.9 atom % 7Li
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Xylitol
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)